Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, suitable for electrophoresis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, meets USP testing specifications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate, 96%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sign Into View Organizational & Contract Pricing Supelco
Dichloromethane, ≥99.9%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sign Into View Organizational & Contract Pricing Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sign Into View Organizational & Contract Pricing