MilliporeSigma
  • EphB4 Expressing Stromal Cells Exhibit an Enhanced Capacity for Hematopoietic Stem Cell Maintenance.
EphB4 Expressing Stromal Cells Exhibit an Enhanced Capacity for Hematopoietic Stem Cell Maintenance.
Stem cells (Dayton, Ohio) (2015-06-03)
Thao M Nguyen, Agnieszka Arthur, Romana Panagopoulos, Sharon Paton, John D Hayball, Andrew C W Zannettino, Louise E Purton, Koichi Matsuo, Stan Gronthos
ABSTRACT
The tyrosine kinase receptor, EphB4, mediates cross-talk between stromal and hematopoietic populations during bone remodeling, fracture repair and arthritis, through its interactions with the ligand, ephrin-B2. This study demonstrated that transgenic EphB4 mice (EphB4 Tg), over-expressing EphB4 under the control of collagen type-1 promoter, exhibited higher frequencies of osteogenic cells and hematopoietic stem/progenitor cells (HSC), correlating with a higher frequency of long-term culture-initiating cells (LTC-IC), compared with wild type (WT) mice. EphB4 Tg stromal feeder layers displayed a greater capacity to support LTC-IC in vitro, where blocking EphB4/ephrin-B2 interactions decreased LTC-IC output. Similarly, short hairpin RNA-mediated EphB4 knockdown in human bone marrow stromal cells reduced their ability to support high ephrin-B2 expressing CD34(+) HSC in LTC-IC cultures. Notably, irradiated EphB4 Tg mouse recipients displayed enhanced bone marrow reconstitution capacity and enhanced homing efficiency of transplanted donor hematopoietic stem/progenitor cells relative to WT controls. Studies examining the expression of hematopoietic supportive factors produced by stromal cells indicated that CXCL12, Angiopoietin-1, IL-6, FLT-3 ligand, and osteopontin expression were more highly expressed in EphB4 Tg stromal cells compared with WT controls. These findings indicate that EphB4 facilitates stromal-mediated support of hematopoiesis, and constitute a novel component of the HSC niche.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ephb4
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
MISSION® esiRNA, targeting human EPHB4
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)