Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, Absolute - Acetone free
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, NMR reference standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, SAJ special grade
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Leptomycin B solution from Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sign Into View Organizational & Contract Pricing