MilliporeSigma
  • Sol-gel synthesis of quaternary (P2O5)55-(CaO)25-(Na2O)(20-x)-(TiO2) x bioresorbable glasses for bone tissue engineering applications (x = 0, 5, 10, or 15).
Sol-gel synthesis of quaternary (P2O5)55-(CaO)25-(Na2O)(20-x)-(TiO2) x bioresorbable glasses for bone tissue engineering applications (x = 0, 5, 10, or 15).
Biomedical materials (Bristol, England) (2015-08-27)
Farzad Foroutan, Nick J Walters, Gareth J Owens, Nicola J Mordan, Hae-Won Kim, Nora H de Leeuw, Jonathan C Knowles
ABSTRACT
In the present study, we report a new and facile sol-gel synthesis of phosphate-based glasses with the general formula of (P2O5)55-(CaO)25-(Na2O)(20-x)-(TiO2) x , where x = 0, 5, 10 or 15, for bone tissue engineering applications. The sol-gel synthesis method allows greater control over glass morphology at relatively low processing temperature (200 °C) in comparison with phosphate-based melt-derived glasses (~1000 °C). The glasses were analyzed using several characterization techniques, including x-ray diffraction (XRD), (31)P magic angle spinning nuclear magnetic resonance ((31)P MAS-NMR), Fourier transform infrared (FTIR) spectroscopy and energy-dispersive x-ray (EDX) spectroscopy, which confirmed the amorphous and glassy nature of the prepared samples. Degradation was assessed by measuring the ion release and pH change of the storage medium. Cytocompatibility was also confirmed by culturing osteoblast-like osteosarcoma cell line MG-63 on the glass microparticles over a seven-day period. Cell attachment to the particles was imaged using scanning electron microscopy (SEM) and confocal laser scanning microscopy (CLSM). The results revealed the potential of phosphate-based sol-gel derived glasses containing 5 or 10 mol% TiO2, with high surface area, ideal dissolution rate for cell attachment and easily metabolized dissolution products, for bone tissue engineering applications.
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
2-Methoxyethanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Formamide, SAJ first grade, ≥98.5%
Sigma-Aldrich
2-Methoxyethanol, JIS special grade, ≥99.0%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Titanium(IV) isopropoxide, packaged for use in deposition systems
Sigma-Aldrich
Formamide, JIS special grade, ≥98.5%
Sigma-Aldrich
Titanium(IV) isopropoxide, 99.999% trace metals basis
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Titanium(IV) isopropoxide, 97%
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Titanium(IV) isopropoxide, ≥97.0%
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%