MilliporeSigma
  • RP-HPLC-DAD-MS(n) Analysis and Butyrylcholinesterase Inhibitory Activity of Barbacenia blanchetii Extracts.
RP-HPLC-DAD-MS(n) Analysis and Butyrylcholinesterase Inhibitory Activity of Barbacenia blanchetii Extracts.
Natural product communications (2015-07-23)
Jósquia S Barbosa, Verônica M Almeida, Rosilene M Marçal, Alexsandro Branco
ABSTRACT
High-performance liquid chromatography (HPLC) coupled to a diode-array detector (DAD) and electrospray ionization mass spectrometry (MS) was applied for the separation and identification of phenolic compounds in the n-hexane, ethyl acetate and methanol crude extract of Barbacenia blanchetii. The MS, MSn and UV data together with chemosystematic evidence allowed the structural characterization of five compounds: tricin, chrysoeriol, epi-gallocatechin, kaempferol 3-O-glucoside and caffeoylquinic acid. At the same time, these extracts were evaluated against butyrylcholinesterase using Ellman's method. All extracts inhibited BuChE in a concentration-dependent form; however, the methanol extract showed a better effect that the other extracts. These compounds have been identified previously in the Velloziaceae family, but for the first time as constituents of B. blanchetii.
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Trizma® hydrochloride, BioXtra, pH 3.5-5.0 (0.5 M in H2O), ≥99.0% (titration)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Trizma® hydrochloride, BioPerformance Certified, suitable for cell culture, ≥99.0% (titration)
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline
Sigma-Aldrich
Trizma® hydrochloride, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Trizma® hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG