MilliporeSigma
  • Elevated mean neutrophil volume represents altered neutrophil composition and reflects damage after myocardial infarction.
Elevated mean neutrophil volume represents altered neutrophil composition and reflects damage after myocardial infarction.
Basic research in cardiology (2015-10-16)
G P J van Hout, W W van Solinge, C M Gijsberts, M P J Teuben, P H C Leliefeld, M Heeres, F Nijhoff, S de Jong, L Bosch, S C A de Jager, A Huisman, P R Stella, G Pasterkamp, L J Koenderman, I E Hoefer
ABSTRACT
Myocardial infarction (MI) induces an inflammatory response in which neutrophils fulfill a prominent role. Mean neutrophil volume (MNV) represents the average size of the circulating neutrophil population. Our goal was to determine the effect of MI on MNV and investigate the mechanisms behind MNV elevation. MNV of 84 MI patients was compared with the MNV of 209 stable angina patients and correlated to simultaneously measured CK levels. Fourteen pigs were subjected to temporary coronary balloon occlusion and blood was sampled at multiple time points to measure MNV. Echocardiography was performed followed by ex vivo infarct size assessment after 72 h. MNV was higher in MI patients compared to stable angina patients (602 SD26 AU vs. 580 SD20 AU, p < 0.0001) and correlated with simultaneously measured CK levels (R = 0.357, p < 0.0001). In pigs, MNV was elevated post-MI (451 SD11 AU vs. 469 SD12 AU), p < 0.0001). MNV correlated with infarct size (R = 0.705, p = 0.007) and inversely correlated with left ventricular ejection fraction (R = -0.718, p = 0.009). Cell sorting revealed an increased presence of banded neutrophils after MI, which have a higher MNV compared to mature neutrophils post-MI (495 SD14 AU vs. 478 SD11 AU, p = 0.012). MNV from coronary sinus blood was higher than MNV of neutrophils from simultaneously sampled arterial blood (463 SD7.6 AU vs. 461 SD8.6 AU, p = 0.013) post-MI. The current study shows MNV is elevated and reflects cardiac damage post-MI. MNV increases due to altered neutrophil composition and systemic neutrophil activation. MNV may be an interesting parameter for prognostic assessment in MI and provide new insights into pathological innate immune responses evoked by ischemia-reperfusion.
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Titanium(IV) chloride solution, 0.09 M in 20% HCl
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, CP
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminum oxide, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Titanium(IV) chloride, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Aluminum oxide, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Titanium(IV) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, Brockmann I, activated, standard grade, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, neutral, free-flowing, Redi-Dri