MilliporeSigma
Prazosin Reduces Alcohol Intake in an Animal Model of Alcohol Relapse.
Alcoholism, clinical and experimental research (2015-07-26)
Janice C Froehlich, Brett Hausauer, Stephen Fischer, Bradley Wise, Dennis D Rasmussen
ABSTRACT
Many alcoholics and heavy drinkers undergo repeated cycles of alcohol abstinence followed by relapse to alcohol drinking; a pattern that contributes to escalated alcohol intake over time. In rodents, alcohol drinking that is interspersed with periods of alcohol deprivation (imposed abstinence) increases alcohol intake during reaccess to alcohol. This is termed the "alcohol deprivation effect" or "ADE" and is a model of alcohol relapse in humans. We have previously reported that prazosin reduces alcohol drinking during both brief and prolonged treatment in rats selectively bred for alcohol preference ("P" rats). This study explores whether prazosin prevents alcohol "relapse" in P rats, as reflected by a reduced or abolished ADE. Adult male P rats were given 24-hour access to food and water and scheduled access to alcohol (15 and 30% v/v solutions presented concurrently) for 2 h/d. After 5 weeks, rats underwent imposed alcohol deprivation for 2 weeks, followed by alcohol reaccess for 2 weeks, and this pattern was repeated for a total of 3 cycles. Rats were injected with prazosin (0, 0.5, 1.0, or 2.0 mg/kg body weight, intraperitoneally) once a day for the first 5 days of each alcohol reaccess cycle. Alcohol intake increased on the first day of each alcohol reaccess cycle, demonstrating the formation of an ADE. The ADE was short-lived, lasting only 1 day, during each of the 3 cycles. Prazosin, in all doses tested, prevented the expression of an ADE in all 3 alcohol reaccess cycles. Prazosin decreases alcohol intake in P rats even in a situation that would be expected to increase alcohol drinking, namely following periods of alcohol deprivation. This suggests that prazosin may be effective in reducing alcohol relapse that often occurs during attempts to achieve permanent alcohol abstinence in treatment-seeking alcoholics and heavy drinkers.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Prazosin hydrochloride, ≥99.0% (HPLC)