MilliporeSigma
  • Ion-pair reversed phase liquid chromatography with ultraviolet detection for analysis of ultraviolet transparent cations.
Ion-pair reversed phase liquid chromatography with ultraviolet detection for analysis of ultraviolet transparent cations.
Journal of chromatography. A (2015-07-22)
Yan He, Kenneth S Cook, Eric Littlepage, John Cundy, Ratish Mangalathillam, Michael T Jones
ABSTRACT
This paper describes the use of an anionic ion-pair reagent (IPR) to impove the ultraviolet (UV) detection and hydrophobic retention of polar and UV transparent cations. Anionic IPR added to the mobile phase forms an ion-pair with cations. Formation of the ion-pair causes a redshift in the absorption wavength, making it possible for direct UV detection of UV-inactive cations. The ion-pairs with increased hydrophobicity were separated by reversed phase liquid chromatography (RPLC). Different perfluorinated caboxylic acids (trifluoroacetic acid, heptafluorobutyric acid, nonafluoropentanoic acid) were evaluted as IPR in the separation and detection of the common cations sodium, ammonium and Tris(hydroxymethyl)aminomethane (Tris). The effects of the IPR type and concentration on separation and detection have been investigated to understand the separation and detection mechanisms. The optimal separation and detection condtions were attained with mobile phase containing 0.1% nonafluoropentanoic acid and with the UV detection at 210nm. UV detection and charged aerosol detection (CAD) were compared in the quantitation of the cations. The limit of quantitation (LOQ) of sodium and Tris with UV detection is comparable to that by CAD. The LOQ of ammonium with UV detection (1ppm or 3ng) is about 20-fold lower than that (20ppm or 60ng) by CAD. The RPLC-UV method was used to monitor ammonium clearance during ultrafiltration and diafiltration in the manfucaturing of biopharmceutical drug substance.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%