MilliporeSigma
  • Versatile (bio)functionalization of bromo-terminated phosphonate-modified porous aluminum oxide.
Versatile (bio)functionalization of bromo-terminated phosphonate-modified porous aluminum oxide.
Langmuir : the ACS journal of surfaces and colloids (2015-04-29)
Aline Debrassi, Esther Roeven, Selina Thijssen, Luc Scheres, Willem M de Vos, Tom Wennekes, Han Zuilhof
ABSTRACT
Porous aluminum oxide (PAO) is a nanoporous material used for various (bio)technological applications, and tailoring its surface properties via covalent modification is a way to expand and refine its application. Specific and complex chemical modification of the PAO surface requires a stepwise approach in which a secondary reaction on a stable initial modification is necessary to achieve the desired terminal molecular architecture and reactivity. We here show that the straightforward initial modification of the bare PAO surface with bromo-terminated phosphonic acid allows for the subsequent preparation of PAO with a wide scope of terminal reactive groups, making it suitable for (bio)functionalization. Starting from the initial bromo-terminated PAO, we prepared PAO surfaces presenting various terminal functional groups, such as azide, alkyne, alkene, thiol, isothiocyanate, and N-hydroxysuccinimide (NHS). We also show that this wide scope of easily accessible tailored reactive PAO surfaces can be used for subsequent modification with (bio)molecules, including carbohydrate derivatives and fluorescently labeled proteins.
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Pricing and availability is not currently available.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Pricing and availability is not currently available.
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Supelco
Dichloromethane, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Pricing and availability is not currently available.