MilliporeSigma
  • Bile acid promotes liver regeneration via farnesoid X receptor signaling pathways in rats.
Bile acid promotes liver regeneration via farnesoid X receptor signaling pathways in rats.
Molecular medicine reports (2015-01-31)
Long Ding, Yu Yang, Yikun Qu, Ting Yang, Kaifeng Wang, Weixin Liu, Weibin Xia
ABSTRACT
Bile acids, which are synthesized from cholesterol in the hepatocytes of the liver, are amphipathic molecules with a steroid backbone. Studies have shown that bile acid exhibits important effects on liver regeneration. However, the mechanism underlying these effects remains unclear. The aim of the present study was to investigate the effect of bile acid and the farnesoid X receptor (FXR) on hepatic regeneration and lipid metabolism. Rats were fed with 0.2% bile acid or glucose for 7 days and then subjected to a 50 or 70% hepatectomy. Hepatic regeneration rate, serum and liver levels of bile acid, and expression of FXR and Caveolin‑1, were detected at 24, 48 or 72 h following hepatectomy. The expression of proliferating cell nuclear antigen (PCNA) in the liver was measured using immunohistochemistry at the end of the study. Hepatocytes isolated from rats were treated with bile acid, glucose, FXR agonist and FXR antagonist, separately or in combination. Lipid metabolism, the expression of members of the FXR signaling pathway and energy metabolism‑related factors were measured using ELISA kits or western blotting. Bile acid significantly increased the hepatic regeneration rate and the expression of FXR, Caveolin‑1 and PCNA. Levels of total cholesterol and high density lipoprotein were increased in bile acid‑ or FXR agonist‑treated hepatocytes in vitro. Levels of triglyceride, low density lipoprotein and free fatty acid were decreased. In addition, bile acid and FXR agonists increased the expression of bile salt export pump and small heterodimer partner, and downregulated the expression of apical sodium‑dependent bile acid transporter, Na+/taurocholate cotransporting polypeptide and cholesterol 7α‑hydroxylase. These results suggested that physiological concentrations of bile acid may promote liver regeneration via FXR signaling pathways, and may be associated with energy metabolism.
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for molecular biology, sterile filtered
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
E-Toxate Water, endotoxin, free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, BioPerformance Certified
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
GW4064, ≥97% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hematoxylin
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloral hydrate, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloral hydrate
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloral hydrate, SAJ special grade, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chloral hydrate, SAJ first grade, ≥99.5%