MilliporeSigma
  • Optical properties of plasmon-resonant bare and silica-coated nanostars used for cell imaging.
Optical properties of plasmon-resonant bare and silica-coated nanostars used for cell imaging.
Journal of biomedical optics (2015-08-01)
Olga Bibikova, Alexey Popov, Alexander Bykov, Artur Prilepskii, Matti Kinnunen, Krisztian Kordas, Vladimir Bogatyrev, Nikolai Khlebtsov, Seppo Vainio, Valery Tuchin
ABSTRACT
We synthesized and characterized gold nanostars and their silica-coated derivatives with 7- to 50-nm shell thicknesses as contrast agents for optical imaging. The scattering and absorption coefficients of the nanoparticles (NPs) were estimated by means of collimated transmittance and diffuse reflectance/transmittance analyses. The contrasting properties of the nanostructures were studied in optical coherence tomography glass capillary imaging. The silica-coated nanostars with the thickest shell have higher scattering ability in comparison with bare nanostars. Viability assays confirmed weak in vitro toxicity of nanostructures at up to ∼200-μg/mL concentrations. We showed real-time visualization of nanostars in both agarose and cultured cells by analyzing the backscattering signal using a conventional laser confocal microscope. The signal intensity detected from the silica-coated NPs was almost 1.5 times higher in comparison with bare nanostars. To the best of our knowledge, this is the first time that conventional laser confocal microscopy was applied in combined scattering and transmitted light modes to detect the backscattered signal of gold nanostars, which is useful for direct monitoring of the uptake, translocation, and accumulation of NPs in living cells.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
VenPure® SF, powder