MilliporeSigma
  • Determination of phthalate esters in environmental water by magnetic Zeolitic Imidazolate Framework-8 solid-phase extraction coupled with high-performance liquid chromatography.
Determination of phthalate esters in environmental water by magnetic Zeolitic Imidazolate Framework-8 solid-phase extraction coupled with high-performance liquid chromatography.
Journal of chromatography. A (2015-08-01)
Xiaomei Liu, Zhiwei Sun, Guang Chen, Wanwan Zhang, Yiping Cai, Rongmei Kong, Xiaoyan Wang, Yourui Suo, Jinmao You
ABSTRACT
In this study, the magnetic Zeolitic Imidazolate Framework-8 (ZIF-8) microspheres were successfully synthesized and applied as an effective sorbent for preconcentration of several typical phthalate esters (PAEs) from environmental water samples. Firstly, the solvothermal treatment method was used for preparation of Fe3O4 nanoparticles. Then, mercaptoacetic acid (MAA) was served as the functionalized chemical to modify Fe3O4 nanoparticles. The Fe3O4@ZIF-8 core-shell microspheres were synthesized through coating the MAA-capped Fe3O4 nanoparticles with ZIF-8. By coupling magnetic solid-phase extraction (MSPE) with high-performance liquid chromatography (HPLC), a reliable, sensitive and cost-effective method for simultaneous determination of five main PAEs including dimethyl phthalate (DMP), diethyl phthalate (DEP), di-n-butyl phthalate (DBP), benzyl butyl phthalate (BBP), and dioctyl phthalate (DOP) was developed. Good linearity was observed in the range of 1.0-100.0μg/L. The limits of detection (S/N=3) and limits of quantification (S/N=10) were in the range of 0.08-0.24 and 0.3-0.8μg/L, respectively. The relative standard deviations were less than 5.5% and the accuracies of the method for the PAEs were in the range from 85.6% to 103.6%. Finally, the Fe3O4@ZIF-8 was successfully applied for rapid extraction of trace amounts of PAEs in environmental water samples.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Benzyl butyl phthalate, 98%
Sigma-Aldrich
Dess-Martin periodinane, 97%
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Dibutyl phthalate, SAJ special grade, ≥98.0%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Dimethyl phthalate, SAJ special grade, ≥99.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Bis(2-ethylhexyl) phthalate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Toluene, JIS 300, for residue analysis, ≥99.8%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300