MilliporeSigma
  • The Effect of Etoricoxib on Hepatic Ischemia-Reperfusion Injury in Rats.
The Effect of Etoricoxib on Hepatic Ischemia-Reperfusion Injury in Rats.
Oxidative medicine and cellular longevity (2015-08-04)
Celalettin Semih Kunak, Osman Kukula, Emre Mutlu, Fatma Genç, Gülçer Güleç Peker, Ufuk Kuyrukluyıldız, Orhan Binici, Durdu Altuner, Hamit Hakan Alp
ABSTRACT
Ischemia-reperfusion (I/R) damage is known to be a pathological process which continues with the increase of oxidants and expands with the inflammatory response. There is not any study about protective effect of etoricoxib on the liver I/R damage in literature. This study investigates the effect of etoricoxib on oxidative stress induced by I/R of the rat liver. Experimental animals were divided into four groups as liver I/R control (LIRC), 50 mg/kg etoricoxib + liver I/R (ETO-50), 100 mg/kg etoricoxib + liver I/R (ETO-100), and healthy group (HG). ETO-50 and ETO-100 groups were administered etoricoxib, while LIRC and HG groups were orally given distilled water by gavage. Hepatic artery was clamped for one hour to provide ischemia, and then reperfusion was provided for 6 hours. Oxidant, antioxidant, and COX-2 gene expressions were studied in the liver tissues. ALT and AST were measured. Etoricoxib in 50 and 100 mg/kg doses changed the levels of oxidant/antioxidant parameters such as MDA, MPO, tGSH, GSHRd, GST, SOD, NO, and 8-OH/Gua in favour of antioxidants. Furthermore, etoricoxib prevented increase of COX-2 gene expression and ALT and AST levels. This important protective effect of etoricoxib on the rat liver I/R can be tested in the clinical setting.
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Phenol, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Phenol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Phenol, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC