MilliporeSigma
Procognitive Compounds Promote Neurite Outgrowth.
Pharmacology (2015-08-01)
Matthew Page, Nathalie Pacico, Sadia Ourtioualous, Tania Deprez, Kyoko Koshibu
ABSTRACT
To this date, the only available drugs for treating Alzheimer's disease are cognitive enhancers, which may improve the cognitive function of patients for a few years while the disease continues to progress. As such, there are intense investigations to develop disease-modifying drugs to suppress progressive neurodegeneration. In this study, a range of procognitive compounds are tested in a primary neuronal culture to determine their relative potential for promoting neuritogenesis. We report that donepezil, memantine, dimebon, Pre-084 and 4-IBP are neuritogenic while tacrine, rosemarinic acid, memoquin and a BACE1 inhibitor suppress neurite outgrowth of neurons. The results of this study indicate that some procognitive compounds may possess a disease-modifying potential.
Sigma-Aldrich
Histamine, ≥97.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 g
In Stock
Details...
$98.70
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 g
In Stock
Details...
$98.70
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 g
In Stock
Details...
$98.70
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 g
In Stock
Details...
$98.70
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 g
In Stock
Details...
$98.70
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 g
In Stock
Details...
$98.70