MilliporeSigma
  • Derivative emission spectrofluorimetry: Application to the analysis of newly approved FDA combination of ibuprofen and famotidine in tablets.
Derivative emission spectrofluorimetry: Application to the analysis of newly approved FDA combination of ibuprofen and famotidine in tablets.
Luminescence : the journal of biological and chemical luminescence (2014-11-26)
Marwa A A Ragab, Eman I El-Kimary
ABSTRACT
A new combination of ibuprofen (NSAID) and famotidine (H2 receptor antagonist) was recently approved by the FDA. It was formulated to relief pain while decreasing the risk of ulceration, which is a common problem for patients receiving NSAID. A rapid and simple derivative emission spectrofluorimetric method is proposed for the simultaneous analysis of this combination in their pharmaceutical preparation. The method is based upon measurement of the native fluorescence intensity of the two drugs at λex = 233 nm in acetonitrile. The emission data were differentiated using the first (D1) derivative technique. The plots of derivative fluorescence intensity versus concentration were rectilinear over a range of 2-35 and 0.4-8 µg/mL for both ibuprofen (IBU) and famotidine (FAM), respectively. The method was sensitive as the limits of detection were 0.51 and 0.12 µg/mL and limits of quantitation were 1.70 and 0.39 µg/mL, for IBU and FAM respectively. The proposed derivative emission spectrofluorimetric method was successfully applied for the determination of the two drugs in their synthetic mixtures and tablets with good accuracy and precision. The proposed method was validated as per ICH guidelines.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M