MilliporeSigma
  • Rapid and MR-Independent IK1 Activation by Aldosterone during Ischemia-Reperfusion.
Rapid and MR-Independent IK1 Activation by Aldosterone during Ischemia-Reperfusion.
PloS one (2015-07-30)
Joachim Alexandre, Thomas Hof, Paolo Emilio Puddu, René Rouet, Romain Guinamard, Alain Manrique, Farzin Beygui, Laurent Sallé, Paul Milliez
ABSTRACT
In ST elevation myocardial infarction (STEMI) context, clinical studies have shown the deleterious effect of high aldosterone levels on ventricular arrhythmia occurrence and cardiac mortality. Previous in vitro reports showed that during ischemia-reperfusion, aldosterone modulates K+ currents involved in the holding of the resting membrane potential (RMP). The aim of this study was to assess the electrophysiological impact of aldosterone on IK1 current during myocardial ischemia-reperfusion. We used an in vitro model of "border zone" using right rabbit ventricle and standard microelectrode technique followed by cell-attached recordings from freshly isolated rabbit ventricular cardiomyocytes. In microelectrode experiments, aldosterone (10 and 100 nmol/L, n=7 respectively) increased the action potential duration (APD) dispersion at 90% between ischemic and normoxic zones (from 95±4 ms to 116±6 ms and 127±5 ms respectively, P<0.05) and reperfusion-induced sustained premature ventricular contractions occurrence (from 2/12 to 5/7 preparations, P<0.05). Conversely, potassium canrenoate 100 nmol/L and RU 28318 1 μmol/l alone did not affect AP parameters and premature ventricular contractions occurrence (except Vmax which was decreased by potassium canrenoate during simulated-ischemia). Furthermore, aldosterone induced a RMP hyperpolarization, evoking an implication of a K+ current involved in the holding of the RMP. Cell-attached recordings showed that aldosterone 10 nmol/L quickly activated (within 6.2±0.4 min) a 30 pS K+-selective current, inward rectifier, with pharmacological and biophysical properties consistent with the IK1 current (NPo =1.9±0.4 in control vs NPo=3.0±0.4, n=10, P<0.05). These deleterious effects persisted in presence of RU 28318, a specific MR antagonist, and were successfully prevented by potassium canrenoate, a non specific MR antagonist, in both microelectrode and patch-clamp recordings, thus indicating a MR-independent IK1 activation. In this ischemia-reperfusion context, aldosterone induced rapid and MR-independent deleterious effects including an arrhythmia substrate (increased APD90 dispersion) and triggered activities (increased premature ventricular contractions occurrence on reperfusion) possibly related to direct IK1 activation.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Calcium chloride
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Calcium chloride solution, 3.2 mM
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Calcium chloride solution, 0.025 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
$233.00