Merck
  • A Comprehensive Systems Biological Study of Autophagy-Apoptosis Crosstalk during Endoplasmic Reticulum Stress.
A Comprehensive Systems Biological Study of Autophagy-Apoptosis Crosstalk during Endoplasmic Reticulum Stress.
BioMed research international (2015-05-20)
Marianna Holczer, Margita Márton, Anita Kurucz, Gábor Bánhegyi, Orsolya Kapuy
ABSTRACT
One of the most important tasks of a living organism is to maintain its genetic integrity with respect to stress. Endoplasmic reticulum (ER) has a crucial role in sensing cellular homeostasis by controlling metabolism, proteostasis, and several signaling processes. ER stressors can induce autophagy-dependent survival; however excessive level of stress results in apoptotic cell death. Although many molecular components of these networks have already been discovered, the analysis of the dynamical features of the regulatory network of life-or-death decision is still lacking. Our goal was to incorporate both theoretical and molecular biological techniques to explore the autophagy-apoptosis crosstalk under ER stress. Using various levels of different ER stressors we confirmed that the control network always generated an evidently detectable autophagy-dependent threshold for apoptosis activation. We explored the features of this threshold by introducing both autophagy activators and inhibitors, and transient treatment with excessive level of ER stressor was also performed. Our experimental data were also supported by a stochastic approach. Our analysis suggests that even if the switch-like characteristic of apoptosis activation is hardly seen on population level the double negative feedback loop between autophagy and apoptosis inducers introduces bistability in the control network.
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Metyrapone, ≥98% (HPLC), solid
SKU
Pack Size
Availability
Price
Quantity
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus