Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Diethanolamine, SAJ first grade, ≥98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Diethanolamine, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri™, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sign Into View Organizational & Contract Pricing Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, meets USP testing specifications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, 99.995% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Sign Into View Organizational & Contract Pricing