MilliporeSigma
UHPLC Analysis of B-vitamins on Titan C18 using UV detection
UHPLC Analysis of B-vitamins on Titan C18 using UV detection application for UHPLC
Materials
standard
Product No.
Description
Pricing
Pyridoxine hydrochloride
meets USP testing specifications
Calcium pantothenate
meets USP testing specifications, monograph mol wt. 476.53 (C18H32CaN2O10)
Folic acid
meets USP testing specifications
Niacinamide
meets USP testing specifications
(−)-Riboflavin
meets USP testing specifications
Biotin
meets USP testing specifications
Cyanocobalamin (B12)
analytical standard
Nicotinic acid
analytical standard
CONDITIONS
column
TITAN C18, 50 mm x 2.1 mm I.D., x 1.9 μm particles (577122-U)
mobile phase
[A] 20 mM potassium phosphate, pH 3; [B] methanol
gradient
0.5% B held for 0.5 minutes, to 30% B in 1.3 minutes, held at 30% B for 1 minute, held at 0.5% B for 0.8 minutes
flow rate
0.5 mL/min
pressure
7977 psi (550 bar)
column temp.
30 °C
detector
UV, 210 nm
injection
0.5 μL
sample
100 ug/mL of all vitamins except 250 ug/mL of calcium pantothenate, biotin and riboflavin in water
Description
Legal Information
null