Merck
HomeBatteries, Supercapacitors & Fuel CellsGC-MS Analysis of Lithium Ion Battery Electrolyte Solvents
GC-MS Determination of Lithium-Ion Battery Electrolyte Solvents and Their Impurities on a SPB®-624 Capillary GC Column
Valanka D’Silva
R&D and Customer Support Lab APAC, Jigani, Bangalore, India
Abstract
A simple and efficient gas chromatography–mass spectrometry method (GC-MS) has been developed for the separation of battery electrolyte solvents used for lithium-ion batteries and their impurities on an SPB®-624 capillary column.
Section Overview:
Introduction
Electrolyte solutions are essential in rechargeable lithium-ion batteries, which are widely utilized in portable electronics and the electric vehicle market.1 These electrolytes facilitate the transport of lithium ions between the cathode and anode, requiring both a high dielectric constant and low viscosity to enable high voltage and specific energy advantages. Commonly used mixtures of linear and cyclic carbonates, such as ethylene carbonate (EC), dimethyl carbonate (DMC), diethyl carbonate (DEC), and ethyl methyl carbonate (EMC), serve this purpose.
The quality of the electrolyte significantly influences battery performance, particularly the presence of trace impurities such as alcohols and esters. These protic impurities, including water, often originate from the solvent manufacturing process or result from degradation reactions. They can react with LiPF6, a frequently used compound in electrolyte formulations, leading to the formation of hydrofluoric acid (HF), which adversely affects electrolyte quality.2 Thus, maintaining the purity of carbonate solvents is crucial for effective electrolyte preparation.
Recent studies have demonstrated that the stability of these electrolyte mixtures can be enhanced by controlling their composition and minimizing impurities.3,4 For instance, the introduction of specific additives has been shown to improve the performance of lithium-ion batteries by forming a stable solid electrolyte interface (SEI) that enhances ion transport while preventing further electrolyte degradation.5 The ongoing research into the optimization of carbonate-based electrolytes is vital for advancing the efficiency and safety of lithium-ion batteries, particularly as the demand for electric vehicles and renewable energy storage solutions continues to grow.
Battery solvent
Common impurities found
Ethylene carbonate (EC)
Dimethyl carbonate (DMC),6 diethyl carbonate (DEC),7 propylene carbonate (PC)8
Dimethyl carbonate (DMC)
Methyl alcohol, formaldehyde,9 methylene chloride,10 methyl formate,11 propylene carbonate12
Diethyl carbonate (DEC)
Dimethyl carbonate (DMC), ethyl alcohol,13 ethylene glycol14
Ethyl methyl carbonate (EMC)
Dimethyl carbonate (DMC), diethyl carbonate (DEC), ethyl alcohol, methyl alcohol9
Vinylene carbonate (VC)
EC, DMC, DEC,15 FEC16
Propylene carbonate (PC)
EC,17 DMC,18 DEC19
Fluoroethylene carbonate (FEC)
EC,20 DMC,21 DEC22
n-Propyl propionate (PP)
Isopropyl propionate,23 n-propyl acetate,24 isopropyl acetate25
Table 1.Common impurities found in battery solvents
Since the presence of trace impurities can significantly impact the efficiency and safety of these batteries, testing the purity of these solvents is of immense importance, ensuring the longevity of battery performance.
The purity of these carbonate solvents can be determined by GC-MS applying a bonded poly(6% cyanopropylphenyl/94% dimethyl siloxane) type capillary column such as the SPB®-624 or OVI-G43, the latter is optimized for determining residual solvents in pharmaceutical preparations. These columns offer the required selectivity for the separation of the carbonate battery solvents and their organic impurities due to their increased phase polarity. In this study here, the SPB®-624 is used.
Experimental
Sample Preparation
Prepare the solvent mix solution as shown in Table 2.
Accurately weigh and transfer A mg of each solvent/impurity into separate 10 mL volumetric flasks. Add 1,4-dioxane and sonicate for 5 minutes. Top up to volume with 1,4-dioxane and mix well. The resulting solutions (1-17) contain B µg/mL of each solvent.
Transfer C mL of each resultant solution into a single 100 mL volumetric flask. Top up to volume with 1,4-dioxane and mix well. The resulting mix solution contains D µg/mL of each solvent.
Solution No.
Solvent
A - Initial weight (mg)
B - Initial solution conc. (µg/mL)
C - Volume of solvent solution (mL)
D - Final solution conc. (µg/mL)
1
Methyl alcohol
80.00
8,000
6.25
500
2
Methyl formate
80.00
8,000
6.25
500
3
Ethyl alcohol
35.00
3,500
5.00
175
4
Methylene chloride
40.00
4,000
5.00
200
5
Formaldehyde
80.00
8,000
6.25
500
6
Dimethyl carbonate
60.00
6,000
5.00
300
7
Isopropyl acetate
20.00
2,000
5.00
100
8
Ethyl methyl carbonate
20.00
2,000
5.00
100
9
n-Propyl acetate
40.00
4,000
5.00
200
10
Isopropyl propionate
20.00
2,000
5.00
100
11
Ethylene glycol
80.00
8,000
5.00
400
12
Diethyl carbonate
20.00
2,000
5.00
100
13
n-Propyl propionate
20.00
2,000
5.00
100
14
Vinylene carbonate
20.00
2,000
5.00
100
15
Fluoroethylene carbonate
30.00
3,000
5.00
150
16
Ethylene carbonate
80.00
8,000
6.25
500
17
Propylene carbonate
40.00
4,000
5.00
200
Table 2.Preparation of solvent mix solution
GC-MS Analysis
The solvent/impurity mixture was analyzed on an SPB®-624 capillary column with conditions stated in Table 3
GC Parameters
Instrument:
Agilent 7890B GC system
Column:
SPB®-624 Capillary GC column, 60 m × 0.32 mm, df 1.8 μm (24251)
Oven:
45 °C for 1 minute, ramped to 200 °C at 10 °C/min and kept for 3 minutes
Inj. Temp.:
200 °C
Carrier gas:
Helium, 2 mL/min
Split ratio:
150:1
Detector:
MSD, EI (see Table 4 for used m/z)
MSD interface:
250 °C
Injection:
1 μL
Liner
4 mm single taper LPD split liner
MS Parameters
Acquisition:
Full scan 13–200 m/z
Collision gas:
Helium
Quench gas:
Nitrogen
Solvent delay:
4 minutes
MS source temperature:
230 °C
Quad temperature:
150 °C
Electron energy:
70 eV
Table 3.GC-MS conditions for analyzing battery solvents and their impurities
Results
The system repeatability is demonstrated with six replicates of solvent mix solution (containing the carbonate battery solvents and their impurities). The separation of the carbonate battery solvents and their impurities is displayed in Figure 1 for the solvent mix solution and the respective chromatographic data is listed in Table 4.
The image is a black-and-white gas chromatography-mass spectrometry (GC-MS) chromatogram depicting the separation of carbonate battery solvents and their impurities in a solvent mix solution. The x-axis represents retention time in minutes, ranging from approximately 4 to 19 minutes, while the y-axis represents intensity in counts per second (cps), ranging from 0 to 1.5 × 10⁸. The chromatogram consists of a series of peaks, each corresponding to different compounds eluted at specific retention times. A major solvent peak, labeled "1,4-dioxane," appears at around 10 minutes, showing the highest intensity. Smaller peaks are distributed across the chromatogram, with retention times ranging from 4.71 minutes (first peak) to 17.91 minutes (last peak). Peaks 1 through 8 appear before the solvent peak, while peaks 10 through 17 elute after it, with peak 14 being notably prominent among them. The chromatogram is plotted with a fine black line on a white background, and the labels for the peaks are written in black text.
Figure 1.GC-MS chromatogram demonstrating the separation of the carbonate battery solvents and their impurities in the solvent mix solution.
Peak no.
Compound
Retention time (min)
% RSD of peak area (n=6)
MDL* (µg/mL)
Mass ions (m/z)
1
Methyl alcohol
4.71
1.38
23.16
29.1, 28.1, 32.1
2
Methyl formate
4.95
1.25
21.07
60.0, 44.1, 45.0
3
Ethyl alcohol
5.53
0.38
2.25
45.1, 46.0, 43.0
4
Methylene chloride
6.61
0.56
3.76
83.9, 85.9, 49.0
5
Formaldehyde
7.59
0.61
10.31
27.8, 31.3, 28.2
6
Dimethyl carbonate
8.19
0.42
4.28
59.0, 45.1, 90.0
7
Isopropyl acetate
8.87
0.74
2.50
43.0, 61.0, 87.0
8
Ethyl methyl carbonate
9.92
0.75
2.52
77.0, 45.1, 59.0
9
n-Propyl acetate
10.02
0.95
6.40
43.0, 61.0, 73.0
10
Isopropyl propionate
10.74
0.60
2.00
57.0, 75.0, 101.1
11
Ethylene glycol
11.17
1.44
3.37
43.0, 61.0, 42.0
12
Diethyl carbonate
11.55
0.36
1.23
91.0, 45.1, 63.0
13
n-Propyl propionate
11.96
0.56
1.88
57.0, 75.0, 87.0
14
Vinylene carbonate
12.32
0.46
1.56
86.0, 42.0, 87.0
15
Fluoroethylene carbonate
15.25
0.29
1.48
62.0, 106.0, 47.0
16
Ethylene carbonate
17.67
1.04
17.46
88.0, 43.0, 44.0
17
Propylene carbonate
17.91
0.39
2.65
57.0, 43.0, 87.0
*MDL=t(n-1, 0.99) SD
Table 4.Chromatographic data of solvent mix solution and typical mass ions
Conclusion
The development of a gas chromatography-mass spectrometry (GC-MS) method using the bonded poly(6% cyanopropyl phenyl/94% dimethyl siloxane) type capillary column SPB®-624 has proven to be effective for the separation and analysis of battery solvents and their known impurities. This method demonstrated excellent selectivity for various carbonate solvents commonly used in lithium-ion batteries, allowing for efficient resolution of the investigated range of analytes within 18 minutes. The full scan acquisition capability of GC-MS facilitates accurate qualitative analysis of typical impurities that may arise during the manufacturing, packaging, or distribution processes of these solvents. This approach not only enhances the understanding of solvent purity but also supports the ongoing efforts to improve the performance and safety of lithium-ion batteries.
Related Products
GC Columns & Accessories
Loading
Reference Materials & Solvents
Loading
References
1.
Manthiram A. 2017. An Outlook on Lithium Ion Battery Technology. ACS Cent. Sci. 3(10):1063-1069. https://doi.org/10.1021/acscentsci.7b00288
2.
Cao X. Important factors for the reliable and reproducible preparation of non-aqueous electrolyte solutions for lithium batteries. Commun Mater. 4(1): https://doi.org/10.1038/s43246-023-00338-7
3.
Aurbach D, Gamolsky K, Markovsky B, Gofer Y, Schmidt M, Heider U. 2002. On the use of vinylene carbonate (VC) as an additive to electrolyte solutions for Li-ion batteries. Electrochimica Acta. 47(9):1423-1439. https://doi.org/10.1016/s0013-4686(01)00858-1
4.
Markevich E, Salitra G, Afri M, Talyosef Y, Aurbach D. 2020. Improved Performance of Li-metal∣LiNi0.8Co0.1Mn0.1O2 Cells with High-Loading Cathodes and Small Amounts of Electrolyte Solutions Containing Fluorinated Carbonates at 30 °C–55 °C. J. Electrochem. Soc. 167(7):070509. https://doi.org/10.1149/1945-7111/ab67a1
5.
Campion CL, Li W, Lucht BL. 2005. Thermal Decomposition of LiPF[sub 6]-Based Electrolytes for Lithium-Ion Batteries. J. Electrochem. Soc. 152(12):A2327. https://doi.org/10.1149/1.2083267
6.
Zhang X, Wang Q, Wang F, Xu J, Xue B. 2024. Efficient Synthesis of Dimethyl Carbonate via Transesterification of Ethylene Carbonate Catalyzed by CN-SiAlO Composites. Catal Lett. 154(10):5416-5428. https://doi.org/10.1007/s10562-024-04739-2
7.
Wang F, Varenne F, Ortiz D, Pinzio V, Mostafavi M, Le Caër S. 2017. Degradation of an Ethylene Carbonate/Diethyl Carbonate Mixture by Using Ionizing Radiation. ChemPhysChem. 18(19):2799-2806. https://doi.org/10.1002/cphc.201700320
8.
Pescarmona PP. 2021. Cyclic carbonates synthesised from CO2: Applications, challenges and recent research trends. Current Opinion in Green and Sustainable Chemistry. 29100457. https://doi.org/10.1016/j.cogsc.2021.100457
9.
Fernandes Y, Bry A, de Persis S. 2019. Thermal degradation analyses of carbonate solvents used in Li-ion batteries. Journal of Power Sources. 414250-261. https://doi.org/10.1016/j.jpowsour.2018.12.077
10.
Zhang M, Xu Y, Williams BL, Xiao M, Wang S, Han D, Sun L, Meng Y. 2021. Catalytic materials for direct synthesis of dimethyl carbonate (DMC) from CO2. Journal of Cleaner Production. 279123344. https://doi.org/10.1016/j.jclepro.2020.123344
11.
Fujinuma N, Page N, Boddy AG, Rivkind J, Tomlinson L, Hoy EP, Lofland SE. 2025. Synergistic Pd‐Au Catalyst for Selective Electrosynthesis of Dimethyl Carbonate in Conjunction with High‐Rate Redox System. Adv Funct Materials. 35(2): https://doi.org/10.1002/adfm.202412402
12.
Liu M, Konstantinova M, Negahdar L, McGregor J. 2021. The role of Zn in the sustainable one-pot synthesis of dimethyl carbonate from carbon dioxide, methanol and propylene oxide. Chemical Engineering Science. 231116267. https://doi.org/10.1016/j.ces.2020.116267
13.
Murugan C, Bajaj H. 2011. Synthesis of diethyl carbonate from dimethyl carbonate and ethanol using KF/Al2O3 as an efficient solid base catalyst. Fuel Processing Technology. 92(1):77-82. https://doi.org/10.1016/j.fuproc.2010.08.023
14.
Kim D, Lee M, Shin Y, Lee J, Lee JW. 2023. Direct production of diethyl carbonate from ethylene carbonate and ethanol by energy-efficient intensification of reaction and separation. Chemical Engineering and Processing - Process Intensification. 192109519. https://doi.org/10.1016/j.cep.2023.109519
15.
Onida K, Ibrahimli L, Duguet N. 2022. Direct Synthesis of Vinylene Carbonates from Aromatic Aldehydes**. Eur J Org Chem. 2022(29): https://doi.org/10.1002/ejoc.202200153
16.
Michan AL, Parimalam BS, Leskes M, Kerber RN, Yoon T, Grey CP, Lucht BL. 2016. Fluoroethylene Carbonate and Vinylene Carbonate Reduction: Understanding Lithium-Ion Battery Electrolyte Additives and Solid Electrolyte Interphase Formation. Chem. Mater. 28(22):8149-8159. https://doi.org/10.1021/acs.chemmater.6b02282
17.
Zhang J, Yang J, Yang L, Lu H, Liu H, Zheng B. Exploring the redox decomposition of ethylene carbonate–propylene carbonate in Li-ion batteries. Mater. Adv. 2(5):1747-1751. https://doi.org/10.1039/d0ma00847h
18.
Shi L, Wang S, Wong DS, Huang K. 2017. Novel Process Design of Synthesizing Propylene Carbonate for Dimethyl Carbonate Production by Indirect Alcoholysis of Urea. Ind. Eng. Chem. Res. 56(40):11531-11544. https://doi.org/10.1021/acs.iecr.7b02341
19.
Shukla K, Srivastava VC. 2018. Efficient Synthesis of Diethyl Carbonate from Propylene Carbonate and Ethanol Using Mg–La Catalysts: Characterization, Parametric, and Thermodynamic Analysis. Ind. Eng. Chem. Res. 57(38):12726-12735. https://doi.org/10.1021/acs.iecr.8b02080
20.
Woo BW, Yoon SW, Lee JH, Park SH, Jang NJ, Yoon HJ; Ulsan Chemical Co. Ltd. Manufacturing method and apparatus of 4-Fluoroethylene carbonate. United States patent US 7,268,238 B2. 2007 Sept 11. . [Internet]. Available from: https://patents.google.com/patent/US7268238B2/en
21.
Yamazaki S, Tatara R, Mizuta H, Kawano K, Yasuno S, Komaba S. 2023. Consumption of Fluoroethylene Carbonate Electrolyte-Additive at the Si–Graphite Negative Electrode in Li and Li-Ion Cells. J. Phys. Chem. C. 127(29):14030-14040. https://doi.org/10.1021/acs.jpcc.3c00843
22.
Jote BA, Beyene TT, Sahalie NA, Weret MA, Olbassa BW, Wondimkun ZT, Berhe GB, Huang C, Su W, Hwang BJ. 2020. Effect of diethyl carbonate solvent with fluorinated solvents as electrolyte system for anode free battery. Journal of Power Sources. 461228102. https://doi.org/10.1016/j.jpowsour.2020.228102
23.
Kastratović V, Radulović M, Kastratović K. 2022. Esterification of propanoic acid in the presence of a homogeneous catalyst. Kragujevac J Science.(44):45-55. https://doi.org/10.5937/kgjsci2244045k
24.
Mitran G, Pavel OD. 2015. Kinetics of acetic acid esterification with propanol in the presence of supported molybdena catalysts. Reac Kinet Mech Cat. 114(1):197-209. https://doi.org/10.1007/s11144-014-0781-4
25.
Guo Y, Zou Y, Huang K, Li Q, Tong Z. 2023. Vapor–Liquid and Chemical Equilibrium for Esterification of Acetic Acid + Isopropanol with [HSO3-bmim][HSO4] at 101.33 kPa. J Solution Chem. 52(11):1209-1231. https://doi.org/10.1007/s10953-023-01311-1