ADC manufacturing: Conjugation
A key step in antibody-drug conjugate (ADC) production is the conjugation of a highly potent active pharmaceutical ingredient to a monoclonal antibody (mAb) using a linker. When conjugated to a targeting antibody, cytotoxic HPAPIs exhibit more selective therapeutic benefits and improved safety profiles.
The linker reacts with accessible lysine or cysteine residues on the surface of the mAb and can result in ADC heterogeneity. Second generation ADCs use engineered mAbs, designed for more uniform linker binding, which reduce variability and increase ADC homogeneity.
Contact us if you need more information on raw materials or equipment for your in-house ADC development or production, or if you need to outsource to a CDMO.
Related Services
- ADC & Bioconjugation CDMO Services
Millipore® CDMO Services excels in antibody-drug conjugate manufacturing.
- Highly Potent APIs
HPAPI contract manufacturing supports drug development with global experience and single-point project management.
- ADC Characterization
Characterization partner: Expert guidance on antibody drug conjugate biological components for safer, reliable testing.
- Extractables and Leachables
Proving that extractables and leachables from your process aren’t harmful to the final drug product is easier with Millipore® Validation Services. Our extractables and leachables (E&L) services thoroughly evaluate all contact materials and processes to mitigate concerns about process fluids.
Development
Initial development of ADC candidates includes the following elements:
- The antibody, which can be a mAb, a bispecific antibody (BsAb), an antigen binding fragment (Fab), or another format.
- A variety of linkers to help maintain stability of the ADC, offer the desired release kinetics, and prevent payload loss, following administration to the patient.
- Different classes of payload that could include small molecules, protein toxins, biologically active peptides, enzymes, and radionuclides.
- Multiple ADC solubilizers, needed to achieve a high drug-antibody ratio (DAR), a wider therapeutic index, and improved drug efficacy.
An efficient conjugation step and generation of multiple ADC candidates is essential to meet development timelines and identify the preferred options for advancing into preclinical and clinical development.
From the earliest stages of development, ADC production should be designed with scalability in mind. Choosing appropriate technologies and equipment that enable easy scaling from bench to large scale will facilitate the transition to clinical- and commercial-scale GMP production. Similarly, containment should be primary focus from the start, as the highly cytotoxic HPAPIs need to be contained to protect operators and the production environment from potential contamination.
Manufacturing
Throughout ADC development and manufacturing, single-use technologies can be used to enhance process flexibility and eliminate the risk of cross-contamination. However, before implementation, it is crucial to confirm that any single-use technology is chemically compatible with the organic solvents used in ADC conjugation. The risk to the ADC from extractables and leachables (E&L) must be considered as they can alter safety, identity, strength, quality, and purity of the drug product. A data-driven, risk-based approach is recommended to support E&L risk assessment efforts and regulatory compliance.
Additional considerations during ADC manufacturing and large-scale conjugation include:
- Ensure robust supply and reliable lead times for raw materials, including the mAb and linker-payload to maintain business continuity and preventing process disruptions.
- Increase the likelihood of successful conjugation by using strong, leak proof containers.
- Improve conjugation efficiency and process robustness through consistent mixing, monitoring and control of key process parameters including temperature and pH.
- Consider ergonomics in containment strategy and equipment selection to enable precise and safe addition of reagents while minimizing drip formation that could compromise the mAb and ADC quality.
Explore our solutions designed to address the challenges of ADC conjugation throughout development and manufacturing:
- High-quality chemicals, essential raw materials and excipients for manufacturing and formulation such as buffers, pH adjusters, solvents, surfactants, and stabilizers. More than 350 chemicals are supported by extensive documentation through our Emprove® Program, accelerating your risk assessment.
- Single-use technologies including Mobius® ADC Reactor and single-use equipment and consumables for TFF, chromatography and aseptic processing to meet different downstream demands in both small- and large-scale processes.
- Real-time monitoring and control solutions, sampling systems, and advanced automation and analytics software, which take process control to a new level.
- Comprehensive contract manufacturing services for mAbs, payloads, linkers, linker-payload intermediates, bioconjugation, and ADC product characterization to accelerate development and manufacturing timelines.
Related Articles
- Leak Resistant Film for Single-Use Bioprocessing Bags
Take advantage of strength and leak prevention in containers made from Ultimus® film. Leak free transportation in single-use bioprocessing bags.
- Mobius® ADC Reactor Single-Use Components Chemical Compatibility
It is important to evaluate the compatibility of the single-use components of an equipment dedicated to conjugation such as the Mobius® ADC reactor.
- Simplifying Antibody-Drug Conjugate Payload Synthesis Using Payload Intermediates
ADC payload intermediates such as, DOLCore ™, MAYCore ™, and PBDCore ™, simplify payload synthesis for the most commonly used payloads: maytansine, dolastatin, and PBD dimers.
- Improving Antibody-drug Conjugate (ADC) Solubility with Hydrophobic Novel Payloads Using the Chito-oligosaccharide ChetoSensar™
See the data on how the chito-oligosaccharide ChetoSensar™ overcomes ADC solubility challenges.
Featured Articles
- Leak Resistant Film for Single-Use Bioprocessing Bags
Take advantage of strength and leak prevention in containers made from Ultimus® film. Leak free transportation in single-use bioprocessing bags.
- Mobius® ADC Reactor Single-Use Components Chemical Compatibility
It is important to evaluate the compatibility of the single-use components of an equipment dedicated to conjugation such as the Mobius® ADC reactor.
- Performance Characteristics of the Mobius® ADC Reactor for Conjugation
ADC production relies on conjugation to link the cytotoxic payload to the mAb. Explore the benefits of a single-use reactor for efficient conjugation.
Web Tools
- Customer Support
Customer support including help with orders, products, accounts, and website technical issues.
- What can we help you with?
Request Information