Cationic Polymers HPLC on TSKgel® PWXL-CP Columns
Conditions | |
---|---|
column | TSKgel G3000PWXL-CP, 30 cm x 7.8 mm, 7 µm particles (821873) |
column | TSKgel G6000PWXL-CP, 30 cm x 7.8 mm, 13 µm particles (821875) |
column | TSKgel G5000PWXL-CP, 30 cm x 7.8 mm, 10 µm particles (821874) |
column temp. | 25 °C |
mobile phase | 0.1 M sodium nitrate |
flow rate | 1 mL/min |
sample | 3 mg/mL |
injection | 100 µL |
detector | refractive index (RI) |
Description | |
---|---|
Analysis Note | Various cationic polymers with different functional groups and molecular weights were injected on the TSKgel PWXL-CP columns (TSKgel G6000PWXL-CP, G5000PWXL-CP and G3000PWXL-CP, connected in series). |
Categories | Analytical Chromatography, Polymers, coatings, and plastics |
Featured Industry | Chemicals and Industrial Polymers |
Other Notes | Data provided by Tosoh Bioscience LLC. |
suitability | application for HPLC |