Merck
HomeSmall Molecules Analysis & QCDetermination of Geniposide Forsythin in XiaoerChiqiaoQingre
HPLC-UV Determination of Geniposide and Forsythin in Xiaoer Chiqiao Qingre Granules acc. to Ch. P. 2020 Method
Dean Duan, Senior Scientist
Merck R&D APAC Lab, Shanghai, China
Abstract
A Discovery® HS C18 HPLC column was used to analyze Xiaoer Chiqiao Qingre granules following the Chinese Pharmacopoeia 2020 monograph. The markers geniposide and forsythin (phillyrin) were analyzed from 1 to 100 and 1 to 200 µg/mL respectively. For the HPLC method, the LOD and LOQ for geniposide are 197.1 and 597.1 mg/g respectively, and for forsythin (phillyrin) 1.5 mg/g and 4.5 mg/g respectively. 
Section Overview
Introduction
Xiaoer Chiqiao Qingre granules is a proprietary traditional Chinese medicine widely used to treat children with symptoms such as fever, cough, nasal congestion, sore throat, and constipation. There are 14 Chinese herbs in this preparation, all of which contain at least 6 active ingredients. Of the active ingredients, only geniposide and forsythin (phillyrin) are used as chemical markers in the Chinese Pharmacopeia 2020 monograph. Here, we use the Discovery® HS C18 column for the HPLC-UV analysis of geniposide and forsythin (phillyrin). The Discovery® HS column is specifically developed for pharmaceutical analysis and purification, showing no detectable bleed in the method and having good lot-to-lot reproducibility. 
The image presents the chemical structures of two compounds: Geniposide and Forsythin. Geniposide, on the left, features a complex multi-ring system with several hydroxyl (OH) groups and a methoxy group (OCH3), displaying both cyclic and acyclic components. Forsythin, on the right, also has a detailed arrangement with multiple rings, hydroxyl groups, and methoxy groups (OCH3).
Experimental
Standard & Mobile Phase Preparation
Geniposide
Mobile phase
  • Take 25 mL of isopropanol and add ~900 mL of water in a 1 L volumetric flask and mix well.
  • Add 0.96 g of citric acid to the solution above and mix well. Top up to mark with water. This is a 5 mM citric acid aqueous solution containing 2.5% isopropanol.
  • To 780 mL of the above, add 220 mL of methanol and mix well. This is the mobile phase for geniposide analysis.
Geniposide standards
  • Weigh ~10 mg (accurate to 0.1 mg) of geniposide into a 10 mL brown volumetric flask.
  • Add ~8 mL of the mobile phase above to it and sonicate for 5 minutes.
  • Top-up to mark with mobile phase and mix well– this is 1 mg/mL of geniposide stock solution.
  • Dilute the geniposide stock solution to 1, 2, 5, 10, 20, 50, and 100 µg/mL with the mobile phase.
Forsythin
Mobile phase
  • Take 220 mL of acetonitrile into a 1 L volumetric flask. Top up with 1 L water and mix well. This is the mobile phase for forsythin analysis.
Forsythin standards
  • Weigh separately ~10 mg (accurate to 0.1 mg) of forsythin (phillyrin) into a 10 mL brown volumetric flask.
  • Add ~8 mL of mobile phase and sonicate for 5 minutes.
  • Top-up to mark with mobile phase and mix well– this is 1 mg/mL of forsynthin stock solution.
  • Dilute to 1, 2, 5, 10, 20, 50, 100, and 200 µg/mL with the mobile phase.
Sample Preparation
Geniposide determination
  • Weigh 0.45 g of ground up Xiaoer Chiqiao Qingre granules into a conical flask.
  • Add 25 mL of methanol to the flask and stopper tightly. Weigh flask and its content.
  • Sonicate for 30 minutes at 300 W/50 kHz and cool to room temperature.
  • Weigh again and use methanol to make up for the lost weight. Mix well and filter.
  • Accurately take 2 mL of the filtrate into a 10 mL volumetric flask. Top up with mobile phase, and mix well for HPLC analysis according to the conditions in Table 2.
Forsythin determination
  • Weigh 2 g of ground up Xiaoer Chiqiao Qingre granules into a conical flask. For a spiked sample for the determination of SPE recovery, add 1 mL of 200 µg/mL standard solution to 2 g of ground granules.
  • Add 25 mL (24 mL for spiked sample) of methanol to the flask and stopper tightly. Weigh the flask and its contents.
  • Sonicate for 30 minutes at 300 W/50 kHz and cool to room temperature.
  • Weigh again and use methanol to make up for the lost weight. Mix well, and filter.
  • Take 10 mL of the filtrate and evaporate to dryness.
  • Add 5 mL of 70% ethanol to dissolve the residue.
  • SPE clean-up of solution according to procedure in Table 1.
  • Analyze according to the conditions in Table 3.
SPE Conditions
SPE tube/cartridge:
Supelclean LC-Alumina-N SPE Tube, 2 g/6 mL (57087)
Conditioning:
5 mL 70% v/v ethanol
Sample addition:
Pre-treated samples for forsythin
Elution:
80 mL of 70% v/v ethanol aqueous solution
Elution post-treatment:
Evaporate to dryness at 45 °C in a water bath. Dissolve residue with 5 mL of 50% v/v aqueous methanol and filter with 0.45 µm filter membrane
Table 1.SPE conditions
LC- Analysis
Prepared samples were analyzed by HPLC-UV according to the two compound specific methods described in Table 2 & 3.
LC Conditions
Column:
Discovery® HS C18, 5 μm, 25 cm × 4.6 mm I.D. (568523-U)
Mobile phase:
[A] 5 mM citric acid aqueous solution with 2.5% v/v isopropanol; [B] methanol
A:B = 90:10
Gradient:
Isocratic
Flow rate:
1.0 mL/min
Pressure:
1590 psi
Column temp.:
35°C
Detector:
UV, 238 nm
Injection:
20 µL
Standards & Samples
Standard:
Geniposide standard solution 20 µg/mL
Sample:
See “Sample Preparation- Geniposide”
Table 2.HPLC Conditions for geniposide determination
LC Conditions
Column:
Discovery® HS C18, 5 μm, 25 cm × 4.6 mm I.D. (568523-U)
Mobile phase:
[A] Water; [B] acetonitrile. A:B = 78:22
Gradient:
Isocratic
Flow rate:
1.0 mL/min
Pressure:
1375 psi
Column temp.:
35°C
Detector:
UV, 277 nm
Injection:
10 µL
Standards & Samples
Standard:
Forsythin standard solution 100 µg/mL
Sample:
See “Sample Preparation - Forsythin”
Table 3.HPLC Conditions for forsythin determination
Method Suitability
Acceptance Criteria: Theoretical plate number for geniposide and forsythin each: NLT 3000
Results
Chromatographic results of standards and samples are displayed in Figure 1–4. The calibration curves for geniposide and forsythin are displayed in Figures 5 and 6, respectively. The specificity of geniposide and forsythin standards are shown in Table 4. The specificity of geniposide and forsythin in Xiaoer Chiqiao Qingre granules is shown in Table 5. Repeatability data is in Table 6, and linearity is in Table 7. The SPE recovery for forsythin is shown in Table 8. The quantification results for the used Xiaoer Chiqiao Qingre granules are shown in Table 8.
See the conclusion for a summary of the results.
HPLC-UV chromatogram of geniposide standard solution with a concentration of 20 μg/mL at 238 nm. Intensity measured in mAU on the y-axis, retention time in minutes on the x-axis. Major ticks on x-axis at 5, 10, 15, 20, and 25 mins; on y-axis at 10, 20, 30, 40, 50, and 60 mAU. Green curve starts at 5 mAU, runs parallel to x-axis, peaks at 10 mins, labeled 1, at 20 mAU, then runs parallel again.
Figure 1.Geniposide (1) standard solution (20 μg/mL).
HPLC-UV chromatogram of a Xiaoer Chiqiao Qingre sample solution treated for the determination of geniposide at 238 nm. Intensity measured in mAU on the y-axis, retention time in minutes on the x-axis. Major ticks on x-axis at 5, 10, 15, 20, and 25 mins; on y-axis at 10, 20, 30, 40, 50, and 60 mAU. Green curve starts at 5 mAU, shows many peaks of different heights, and then a peak, labeled as 1 for geniposide, at 10 minutes corresponding to 26 mAU. Following which there is another small peak and then the curve runs parallel to x-axis again.
Figure 2.Geniposide (1) in Xiaoer Chiqiao Qingre sample solution.
HPLC-UV chromatogram of forsythin standard solution with a concentration of 100 μg/mL at 277 nm. Intensity measured in mAU on the y-axis, retention time in minutes on the x-axis. Major ticks on x-axis at 10, 20, and 30 mins; on y-axis at 10, 20, 30, 40, 50, 60, 70, 80, 90, and 100 mAU. Green curve starts at 5 mAU, runs parallel to x-axis, peaks at 21 mins, labeled 1, at 20 mAU, then runs parallel again.
Figure 3.Forsythin (1) standard solution (100 μg/mL).
HPLC-UV chromatogram of a Xiaoer Chiqiao Qingre sample solution treated for the determination of forsythin at 277 nm. Intensity measured in mAU on the y-axis, retention time in minutes on the x-axis. Major ticks on x-axis at 5, 10, 15, 20, 25, and 30 mins; on y-axis at 10, 20, 30, 40, 50, 60, 70, 80, 90, and 100 mAU. Green curve starts at 5 mAU, shows many peaks of different heights, and then a peak, labeled as 1 for forsythin, at 21 minutes corresponding to 59 mAU. Following which there are other smaller peaks as well.
Figure 4.Forsythin (1) in Xiaoer Chiqiao Qingre sample solution.
Chromatographic Data for Geniposide and Forsythin Analysis
Compound
Retention Time (min)
RRT
Theoretical Plates
Tailing Factor
Geniposide
9.9
1.00
9640
1.01
Forsythin
21.1
2.13
17263
0.97
Table 4.Chromatographic data of geniposide and forsythin standard injections using the two methods in Table 2 and 3.
Compound
Retention Time (min)
Resolution
Theoretical Plates
Tailing Factor
Geniposide
10.1
20.6
9312
1.12
Forsythin
21.1
2.8
14225
0.97
Table 5.Chromatographic data of sample solutions for geniposide and forsythin determinations using the described sample preparations and the two methods in Table 2 and 3.
Sample
Area (µV * s)
Geniposide
(10 µg/mL)
Forsythin
(50 µg/mL)
STD 1
129,827
256,081
STD 2
132,560
259,266
STD 3
130,152
258,932
STD 4
129,587
256,923
STD 5
128,428
257,655
Mean
130,111
257,771
Standard Deviation
1516
1339
RSD (%)
1.16
0.52
Table 6.Repeatability for geniposide and forsythin standards
Compound
Calibration Range (µg/mL)
No. of Calibrators
Linear equation
R2
Granule Sample LOD (µg/g)
Granule Sample
LOQ  (µg/g)
Geniposide
1 - 100
7
y = 14,076.8666x - 14,029.9889
0.9982
197
597
Forsythin
1 - 200
8
y = 13,671.4109x - 9,460.6930
0.9956
9.2
27.7
Table 7.Linearity/calibration data for standard injections and resulting calculated sensitivity for geniposide and forsythin in granule samples
Intensity v/s concentration measured in µg/mL plot representing calibration curve obtained for geniposide at concentrations of 1, 2, 5, 10, 20,50 and 100 µg/mL. Major ticks on x-axis at 10, 20, 30, 40, 50, 60, 70, 80, 90, and 100 µg/mL and on y-axis at 20, 40, 60, 80, 100, 120, 140, and 160 µV*sec × 100000. The linear equation obtained is y = 14,076.8666x - 14,029.9889 with R² = 0.9982.
Figure 5.Calibration curve for standards of geniposide at 1, 2, 5, 10, 20, 50, and 100 µg/mL.
Intensity v/s concentration measured in µg/mL plot representing calibration curve obtained for forsythin at concentrations of 1, 2, 5, 10, 20,50, 100 and 200 µg/mL. Major ticks on x-axis at 20, 40, 60, 80, 100, 120, 140, 160, 180, and 200 µg/mL and on y-axis at 2, 4, 6, 8, 10, 12, and 14 µV*sec × 100000. The linear equation obtained is y = 13,671.4109x - 9,460.6930 with R² = 0.9956.
Figure 6.Calibration curve for standards of forsythin at 1, 2, 5, 10, 20,50, 100 and 200 µg/mL.
Identity
Recovery (%)
Test 1
93.4
Test 2
87.1
Test 3
89.2
Mean
89.9
Standard Deviation
3.2
RSD (%)
3.6
Table 8.SPE recovery for forsythin spiked at 100 µg/g granule sample
Compound
Amount (µg/g)
Geniposide
5510
Forsythin
570
Table 9.Quantification results for Xiaoer Chiqiao Qingre granules used as sample for this study (Figure 2 and 4)
Conclusion
The Discovery® HS C18 column was used for the HPLC-UV determination of geniposide and forsythin (phillyrin) in Xiaoer Chiqiao Qingre granules according to the Chinese Pharmacopeia monograph. The acceptance criteria stated in the monograph were met by the two used methods. The R2 values for the linear range of both geniposide and forsythin calibration were >0.99.
The LOD and LOQ of geniposide were for the standards injections 0.89 and 2.69 µg/mL respectively, representing for the granule samples 197.1 and 597.1 µg/g respectively. The LOD and LOQ of the forsythin (phillyrin) standard were 1.5 and 4.5 µg/ml respectively, representing fro the granules 9.2 and 27.7 µg/g respectively. The Supelclean™ LC-Alumina-N SPE cleanup procedure for forsythin showed a recovery of 89.9% for forsythin.
We have shown that the Discovery® HS C18 column, together with Supelclean LC-Alumina-N SPE can meet the suitability criteria stated in the Chinese Pharmacopeia monograph for the analysis of Xiaoer Chiqiao Qingre granules.
See more applications on Pharmaceutical Analysis & Quality Control.
Related Products
Loading
References
1.
Chinese Pharmacopeia 2020 monograph, volume 1, page 582-583.. [Internet]. Available from: http://down.foodmate.net/standard/index