MilliporeSigma
HomeCell SignalingCyclic Nucleotide-Gated (CNG) and Hyperpolarization Activated Cyclic Nucleotide-Gated (HCN) Channels
Cyclic Nucleotide-Gated (CNG) and Hyperpolarization Activated Cyclic Nucleotide-Gated (HCN) Channels
Cyclic nucleotide-regulated cation channels are classified into two principal subfamilies, the cyclic nucleotide-gated (CNG) channels and the hyperpolarization-activated cyclic nucleotide-gated (HCN) channels. The two channel families differ from each other with regard to their mode of activation. CNG channels are typical ligand-gated channels because their activation requires the binding of cAMP or cGMP. In contrast, HCN channels are principally operated by voltage. These channels open at hyperpolarized membrane potentials and close on depolarization. However, apart from their voltage sensitivity, HCN channels are also activated directly by cyclic nucleotides, which act by shifting voltage dependence of channel activation to more positive potentials.
CNG and HCN channels are members of an extended superfamily of cation channels that is characterized by a principal building unit containing six transmembrane helices (S1-S6) and an ion conducting reentrant pore-loop between S5 and S6. CNG channels conduct calcium and a variety of monovalent cations whereas HCN channels only pass monovalents. Regulation by cyclic nucleotides is conferred by a cyclic nucleotide-binding domain (CNBD) present in the carboxy terminus. Activation of HCN channels by hyperpolarization is controlled by the positively charged S4 helix carrying nine regularly spaced arginine or lysine residues at every third position.
CNG channels are expressed in retinal photoreceptors and olfactory neurons and play a key role in visual and olfactory signal transduction. Defects in CNG channel function cause retinal diseases such as retinitis pigmentosa and total colorblindness (achromatopsia). In addition, CNG channels are found at low density in some other cell types and tissues such as brain, testis and kidney. The physiological role of CNG channels in these tissues is not known yet. In vertebrates the CNG channel family comprises six homologous members. Based on phylogenetic relationship, these proteins are divided into two subfamilies, the A subunits (CNGA1-4) and the B subunits (CNGB1 and CNGB3). Native CNG channels are heterotetramers with different heteromers displaying distinct nucleotide sensitivity, ion selectivity and modulation by calcium. The subunit composition and stoichiometry has been determined for three native channels: the rod and cone photoreceptor channels and the olfactory channel.
Several drugs have been reported to block CNG channels, although not with very high affinity. The most specific among these drugs is L-cis diltiazem which blocks CNG channels in a voltage-dependent manner at micromolar concentrations. High affinity binding of L-cis diltiazem is only seen in heteromeric CNG channels containing the CNGB subunits. CNG channels are also moderately sensitive to blockage by some other inhibitors of the L-type calcium channel (e.g. nifedipine), the local anaesthetic tetracaine and calmodulin antagonists.
HCN channels represent the molecular correlate of the hyperpolarization-activated cation current, Ih. The channels play a central role in the initiation and control of the heart beat. Enhancement of channel activity by binding of cAMP represents the major mechanism by which norepinephrine and other adrenergic agonists increase heart rate. In brain, HCN channels serve to support multiple functions including sleep-wake cycle, motor learning, and dendritic signal integration. The HCN channel family comprises four homologous members (HCN1-4). These subunits assemble to homomeric and heteromeric tetramers with distinct activation thresholds, opening kinetics and responsiveness to cAMP.
There is significant therapeutic potential for drugs that modulate HCN channels. Dysfunction of HCN channels has been linked to cardiac dysrhythmia, ataxia, absence epilepsy, and neuropathic pain syndromes. Blockers of HCN channels have been pursued as potential bradycardiac, antiepileptic and analgetic agents. The most extensively studied blockers of HCN channels are ZD7288 and ivabradine. Both agents block HCN channels in the low micromolar range and reduce heart rate in a variety of species including man.
The Table below contains accepted modulators and additional information.
Abbreviations
W-7: N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride
ZD7288: (N-Ethyl-N-phenylamino)-1,2 dimethyl-6-(methylamino) pyridinium chloride
References
1.
Ashcroft FM. 2000. Cyclic nucleitide-gated channels.199-210. https://doi.org/10.1016/b978-012065310-2/50012-6
2.
Benarroch EE. 2013. HCN channels: Function and clinical implications. Neurology. 80(3):304-310. https://doi.org/10.1212/wnl.0b013e31827dec42
3.
Biel M, Michalakis S. 2007. Function and Dysfunction of CNG Channels: Insights from Channelopathies and Mouse Models. Mol Neurobiol. 35(3):266-277. https://doi.org/10.1007/s12035-007-0025-y
4.
Biel M. 2002. Cardiac HCN Channels Structure, Function, and Modulation. 12(5):206-213. https://doi.org/10.1016/s1050-1738(02)00162-7
5.
Biel M, Seeliger M, Pfeifer A, Kohler K, Gerstner A, Ludwig A, Jaissle G, Fauser S, Zrenner E, Hofmann F. 1999. Selective loss of cone function in mice lacking the cyclic nucleotide-gated channel CNG3. Proceedings of the National Academy of Sciences. 96(13):7553-7557. https://doi.org/10.1073/pnas.96.13.7553
6.
Herrmann S, Hofmann F, Stieber J, Ludwig A. 2012. HCN channels in the heart: lessons from mouse mutants. 166(2):501-509. https://doi.org/10.1111/j.1476-5381.2011.01798.x
7.
Hofmann F, Biel M, Kaupp UB. 2003. International Union of Pharmacology. XLII. Compendium of Voltage-Gated Ion Channels: Cyclic Nucleotide-Modulated Channels. Pharmacol Rev. 55(4):587-589. https://doi.org/10.1124/pr.55.4.10
8.
Kaupp UB, Seifert R. 2002. Cyclic Nucleotide-Gated Ion Channels. Physiological Reviews. 82(3):769-824. https://doi.org/10.1152/physrev.00008.2002
9.
Kaupp UB, Seifert R. 2001. Molecular Diversity of Pacemaker Ion Channels. Annu. Rev. Physiol.. 63(1):235-257. https://doi.org/10.1146/annurev.physiol.63.1.235
10.
Ludwig A, Zong X, Jeglitsch M, Hofmann F, Biel M. 1998. A family of hyperpolarization-activated mammalian cation channels. Nature. 393(6685):587-591. https://doi.org/10.1038/31255
11.
Ludwig A. 2003. Absence epilepsy and sinus dysrhythmia in mice lacking the pacemaker channel HCN2. 22(2):216-224. https://doi.org/10.1093/emboj/cdg032
12.
Matulef K, Zagotta WN. 2003. Cyclic Nucleotide-Gated Ion Channels. Annu. Rev. Cell Dev. Biol.. 19(1):23-44. https://doi.org/10.1146/annurev.cellbio.19.110701.154854
13.
Noam Y, Bernard C, Baram TZ. 2011. Towards an integrated view of HCN channel role in epilepsy. Current Opinion in Neurobiology. 21(6):873-879. https://doi.org/10.1016/j.conb.2011.06.013
14.
Postea O, Biel M. 2011. Exploring HCN channels as novel drug targets. Nat Rev Drug Discov. 10(12):903-914. https://doi.org/10.1038/nrd3576
15.
Reid CA, Phillips AM, Petrou S. 2012. HCN channelopathies: pathophysiology in genetic epilepsy and therapeutic implications. 165(1):49-56. https://doi.org/10.1111/j.1476-5381.2011.01507.x
16.
Robinson RB, Siegelbaum SA. 2003. Hyperpolarization-Activated Cation Currents: From Molecules to Physiological Function. Annu. Rev. Physiol.. 65(1):453-480. https://doi.org/10.1146/annurev.physiol.65.092101.142734
17.
Zagotta WN, Olivier NB, Black KD, Young EC, Olson R, Gouaux E. 2003. Structural basis for modulation and agonist specificity of HCN pacemaker channels. Nature. 425(6954):200-205. https://doi.org/10.1038/nature01922
18.
Zhong H, Molday LL, Molday RS, Yau K. 2002. The heteromeric cyclic nucleotide-gated channel adopts a 3A:1B stoichiometry. Nature. 420(6912):193-198. https://doi.org/10.1038/nature01201