MilliporeSigma
HomeCell SignalingP2 Receptors: P2X Ion Channel Family
P2 Receptors: P2X Ion Channel Family
Once known as P2 purinoreceptors, surface receptors for extracellular nucleotides are now called P2 receptors. This subtle change in nomenclature reflects the more varied nature of nucleotidic ligands, other than those containing a purine moiety, that are capable of activating these surface receptors. The current nomenclature system for P2 receptors is also based on their molecular structure and signal transduction mechanisms, so defining a family of ionotropic P2 receptors (P2X Ligand-Gated Ion Channels, LGICs) and another family of metabotropic P2 receptors (P2Y G Protein-Coupled Receptors, GPCRs).
The seven subunits forming P2X receptors show a common topology of i) intracellular N- and C-termini which possess consensus binding motifs for protein kinases, ii) two transmembrane spanning regions (TM1 and TM2) which line the ion pore, iii) a large extracellular loop, possibly folded into a β-sheet, with 10 conserved cysteine residues forming a series of disulphide bridges, iv) a hydrophobic H5 region close to the pore vestibule, for receptor/channel modulation by extracellular cations (e.g. magnesium, calcium, zinc, copper and hydrogen ions), and v) an ATP-binding site, which involves amino acid residues in the extracellular loop immediately adjacent to TM1 and TM2. The P2X1-7 receptor subunits show 30-50% sequence identity at the peptide level. The stoichiometry of assembled P2X1-7 receptors is thought to involve three subunits, arranged as a stretched trimer.
Triplets of identical P2X subunits form homomeric assemblies and produce the P2X1-7 receptors. Additionally, some P2X receptors exist as heterotrimeric assemblies (e.g. P2X1/2 in SCG neurons, P2X2/3 in sensory ganglia, P2X2/6 and P2X4/6 in CNS neurons, P2X1/5 in some blood vessels). Members of this extended P2X family of homomeric and heteromeric assemblies show many pharmacological and operational differences. For example, the potency orders for nucleotidic agonists vary significantly between P2X receptor subtypes. With blocking agents, some P2X subtypes (P2X4 and P2X4/6) are relatively insensitive to potent antagonists of other P2X receptor subtypes. Agonism and antagonism at some P2X receptors, notably P2X2 and P2X2/3, are affected by extracellular H+ ions. The kinetics of activation, inactivation and deactivation vary considerably among P2X receptor subtypes. Calcium permeability is high for some P2X receptor subtypes, amounting to some 6-10% of the ionic current carried. The P2X7 receptor converts from an ion channel to a pore and, in some cases, this conversion brings about cell death. Other P2X receptors (P2X2, P2X2/3, P2X4 and P2X2/6) show reversible and time-dependent changes in the ion permeability properties of their intrinsic ion channel but here, this phenomenon is not as profound as seen with P2X7.
The loci of the P2X1-7 genes have been defined in the genome of human, rat and, importantly, mice. Some P2X receptors also have been identified from cDNA libraries of chimp, dog, guinea-pig, frog and zebrafish, as well from invertebrates such as Schistosoma mansoni. Notably, the zebrafish genome contains nine P2X genes of which two are paralogues for P2X3 and P2X4 receptors. All P2X genes show a complex organization, with the encoding sequences interrupted by up to 14 introns. Occasionally, errors occur in P2X gene transcription, which yield splice variants that are either dominant negative, modulatory or functionless. Using mice, gene deletion has been achieved for P2X1, P2X2, P2X3 and P2X7 receptors. None are lethal, but P2X1-null homozygote males are infertile. Disruption of either P2X2 or P2X3 genes causes hyporeflexia, whereas knock-out of the P2X7 gene has a profound effect on neuropathic pain.
The Tables below contains accepted modulators and additional information. For a list of additional products, see the Materials section below.
Footnotes
a) P2X1 resembles the P2X receptor in smooth muscle (SM) cells.
b) P2X2 resembles the P2X receptor in neurons (N).
c) P2X7 is the cytolytic P2Z receptor.
d) Length of individual subunits in amino acid residues (aa). Functional channels require three subunits, and all subunits can form functional homotrimeric assemblies in expression systems (Xenopus oocytes, HEK293 cells, CHO cells). Endogenous P2X receptors also can exist as heterotrimeric assemblies comprising two or three different subunits.
e) P2X6 subunits cannot efficiently form a homomultimeric receptor as these subunits are mostly retained in the ER, but they can readily form heteromultimeric ion channels with other subunits (e.g. P2X2/6 and P2X4/6).
f) There are no truly selective agonists for P2X receptors which are all activated by ATP, although each subtype can be distinguished by potency ratios for ATP and two or more nucleotide agonists. Agonist potency is enhanced under acidic conditions at P2X2 receptors, but reduced at P2X1,3,4,7 receptors.
g) There are no truly selective antagonists for P2X subtypes, except for P2X7 which is blocked by KN-62 and hexamethylene amiloride (HMA). Human P2X4 receptors are relatively insensitive to known P2 receptor antagonists, and rat P2X4 even more so, although hP2X4 can be blocked by BBG. Suramin is particularly effective at P2X2 receptors under acidic conditions (pH 5.5).
h) KN-62, KN-04 and HMA are more potent at human than rat P2X7 receptors, and vice versa for BBG.
i) P2X1-7 receptors form intrinsic cation channels that are permeable to Na+, K+ and Ca2+ ions. The permeability properties of some P2X receptors change during prolonged activation. The P2X7 converts from an ion channel to a pore that is permeable to large molecules (400-900 Da). P2X receptor subtypes can be distinguished by their desensitization rates.
j) [35S]ATPγS should be used in the absence of divalent cations.
k) Loss-of-function polymorphism of P2X7 receptors in macrophages abolishes ATP-mediated killing of Mycobacterium tuberculosis.
Abbreviations
A-317491: [3-Phenoxybenzyl-(1,2,3,4-tetrahydro-napthalene-1-yl)-carbamoyl]-benzene-1,2,4-tricarboxylic acid
Ap4A: Diadenosine tetraphosphate
Ap5A: Diadenosine pentaphosphate
ATP: Adenosine 5´-triphosphate
ATPγS: Adenosine 5´-O-(3-thiotriphosphate)
BBG: Brilliant blue G
BzATP: 3'-Benzoylbenzoyl adenosine 5´-triphosphate
CTP: Cytidine 5´-triphosphate
GTP: Guanosine 5´-triphosphate
HMA: Hexamethylene amiloride
HT-AMP: 2-Hexylthioadenosine 5´-monophosphate
Ip5I: Diinosine pentaphosphate
isoPPADS: Pyridoxal-5-phosphate-6-azophenyl-2',5'-disulphonic acid
KN-62: 1-[N,O-bis(5-Isoquinolinesulfonyl)-N-methyl-L-tyrosyl]-4-phenylpiperazine
2-MeSATP: 2-Methylthioadenosine 5'-triphosphate
α,β-MeATP: α,β-Methylene-adenosine-5'-triphosphate
D-β,γ-MeATP: D-β,γ-Methylene-adenosine-5'-triphosphate
L-β,γ-MeATP: L-β,γ-Methylene-adenosine-5'-triphosphate
MRS 2159: Pyridoxal-5-phosphate-6-azophenyl-4'-carboxylate
MRS 2500: (N)-Methanocarba-N6-methyl-2-iodo-2'-deoxyadenosine-3',5'-bisphosphate
NF023: 8,8'-(Carbonylbis(imino-3,1-phenylene carbonylimino)bis(1,3,5-napththalenetrisulfonic acid)
NF279: 8,8'-(Carbonylbis(imino-4,1-phenylenecarbonylimino-4,1-phenylenecarbonylimino)bis(1,3,5-napththalenetrisulfonic acid)
NF449: 8,8'-(Carbonylbis(imino-5,1,3-benzenetriylbis(carbonylimino))tetrakis-benzene-1,3-disulphonic acid)
PAPET-ATP: 2-[2-(4-Aminophenyl)ethylthio]adenosine 5´-triphosphate
Phenol red: Phenolsulfonphthalein sodium salt
Phenolphthalein: 3,3-bis[4-Hydroyphenol]-1(3H)-isofuranone
PPADS: Pyridoxal-5-phosphate-6-azophenyl-2',4'-disulphonic acid
PPNDS: Pyridoxal-5-phosphate-6-(2´-naphthylazo-6-nitro-4´,8´-disulphonate)
RB-2: Reactive blue 2
TNP-ATP: 2',3'-O-(2,4,6-Trinitrophenyl) adenosine triphosphate
UTP: Uridine 5´-triphosphate
Similar Products
Loading
References
1.
Abbracchio MP, Burnstock G. 1994. Purinoceptors: Are there families of P2X and P2Y purinoceptors?. Pharmacology & Therapeutics. 64(3):445-475. https://doi.org/10.1016/0163-7258(94)00048-4
2.
Barrera NP, Ormond SJ, Henderson RM, Murrell-Lagnado RD, Edwardson JM. 2005. Atomic Force Microscopy Imaging Demonstrates that P2X2 Receptors Are Trimers but That P2X6 Receptor Subunits Do Not Oligomerize. J. Biol. Chem.. 280(11):10759-10765. https://doi.org/10.1074/jbc.m412265200
3.
Buell G, Collo G, Rassendren F. 1996. P2X Receptors: An Emerging Channel Family. 8(10):2221-2228. https://doi.org/10.1111/j.1460-9568.1996.tb00745.x
4.
Chessell IP, Hatcher JP, Bountra C, Michel AD, Hughes JP, Green P, Egerton J, Murfin M, Richardson J, Peck WL, et al. 2005. Disruption of the P2X7 purinoceptor gene abolishes chronic inflammatory and neuropathic pain. Pain. 114(3):386-396. https://doi.org/10.1016/j.pain.2005.01.002
5.
Evans RJ. 2009. Orthosteric and allosteric binding sites of P2X receptors. Eur Biophys J. 38(3):319-327. https://doi.org/10.1007/s00249-008-0275-2
6.
Jiang L. 2009. Inhibition of P2X7 receptors by divalent cations: old action and new insight. Eur Biophys J. 38(3):339-346. https://doi.org/10.1007/s00249-008-0315-y
7.
Khakh BS. 2001. Molecular physiology of p2x receptors and atp signalling at synapses. Nat Rev Neurosci. 2(3):165-174. https://doi.org/10.1038/35058521
8.
Kucenas S, Li Z, Cox J, Egan T, Voigt M. 2003. Molecular characterization of the zebrafish P2X receptor subunit gene family. Neuroscience. 121(4):935-945. https://doi.org/10.1016/s0306-4522(03)00566-9
9.
Kucenas S, Li Z, Cox J, Egan T, Voigt M. 2003. Molecular characterization of the zebrafish P2X receptor subunit gene family. Neuroscience. 121(4):935-945. https://doi.org/10.1016/s0306-4522(03)00566-9
10.
Li Z, Liang D, Chen L. 2008. Potential Therapeutic Targets for ATP-Gated P2X Receptor Ion Channels. ASSAY and Drug Development Technologies. 6(2):277-284. https://doi.org/10.1089/adt.2007.121
11.
Mulryan K, Gitterman DP, Lewis CJ, Vial C, Leckie BJ, Cobb AL, Brown JE, Conley EC, Buell G, Pritchard CA, et al. 2000. Reduced vas deferens contraction and male infertility in mice lacking P2X1 receptors. Nature. 403(6765):86-89. https://doi.org/10.1038/47495
12.
Murrell-Lagnado RD, Qureshi OS. 2008. Assembly and trafficking of P2X purinergic receptors (Review). Molecular Membrane Biology. 25(4):321-331. https://doi.org/10.1080/09687680802050385
13.
North RA. 2002. Molecular Physiology of P2X Receptors. Physiological Reviews. 82(4):1013-1067. https://doi.org/10.1152/physrev.00015.2002
14.
Roberts JA, Evans RJ. 2004. ATP Binding at Human P2X1Receptors. J. Biol. Chem.. 279(10):9043-9055. https://doi.org/10.1074/jbc.m308964200
15.
Silberberg SD, Chang T, Swartz KJ. 2005. Secondary Structure and Gating Rearrangements of Transmembrane Segments in Rat P2X4 Receptor Channels. 125(4):347-359. https://doi.org/10.1085/jgp.200409221
16.
Torres GE, Egan TM, Voigt MM. 1999. Hetero-oligomeric Assembly of P2X Receptor Subunits. J. Biol. Chem.. 274(10):6653-6659. https://doi.org/10.1074/jbc.274.10.6653
17.
Yan Z, Liang Z, Tomi? M, Obsil T, Stojilkovic SS. 2005. Molecular Determinants of the Agonist Binding Domain of a P2X Receptor Channel. Mol Pharmacol. 67(4):1078-1088. https://doi.org/10.1124/mol.104.010108