44940
Triple Sugar Iron Agar
NutriSelect® Basic, suitable for microbiology
Synonym(s):
TSI Agar
About This Item
Recommended Products
sterility
non-sterile
Quality Level
product line
BioChemika
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 12 g/L
ferrous sulfate, 0.2 g/L
glucose, 1 g/L
lactose, 10 g/L
meat extract, 3 g/L
mixed peptone, 20 g/L
phenol red, 0.025 g/L
sodium chloride, 5 g/L
sodium thiosulfate, 0.3 g/L
sucrose, 10 g/L
yeast extract, 3 g/L
manufacturer/tradename
NutriSelect® Basic
technique(s)
microbe id | utilization test: suitable
microbiological culture: suitable
final pH
7.4±0.2 (25 °C)
application(s)
clinical testing
environmental
food and beverages
pharmaceutical
water monitoring
microbiology
suitability
nonselective and differential for Citrobacter spp.
nonselective and differential for Escherichia coli
nonselective and differential for Klebsiella spp.
nonselective and differential for Proteus spp.
nonselective and differential for Salmonella spp.
nonselective and differential for Shigella spp.
nonselective and differential for coliforms
nonselective and differential for enterobacteriaceae
Application
Preparation Note
Other Notes
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
44940-100G:
44940-VAR:
44940-500G:
44940-BULK:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Microbiological assessment of bacteria found in spices, a potent source for food spoilage and pathogens.
Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.
An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service