Merck
HS23460
Tough-Tags Label Dispensing Station
別名:
solvent resistant labels, temperature resistant labels
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41106300
NACRES:
NB.22
物質
blue acrylic
W × H × D
280 × 120 × 89 mm
詳細
Organize your lables for easy accessibility. Portable label station holds up to six standard size boxes of Tough-Tags, Cryo-Babies®, and Tough-Spots®. Drop the box in the label station and feed the paper through the slot. Labels are not included and must be purchased separately.
特徴および利点
  • Acrylic station includes a writing surface, a cutting edge for easy dispensing, and non-skid rubber feet
  • counter-sunk holes provided for wall mounting
  • translucent blue
法的情報
Cryo-Babies is a registered trademark of Diversified Biotech
Tough-Spots is a registered trademark of Diversified Biotech
Tough-Tags is a trademark of Diversified Biotech
最新バージョンのいずれかを選択してください:
試験成績書(COA)