주요 문서
HAM7657-01
Hamilton® TLC syringe
model, 1725 RN, no needle, volume 250 μL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
42142600
NACRES:
NB.03
material
Gastight syringe
특징
autoclavable
model 1725 RN, no needle
제조업체/상표
Hamilton
파라미터
10-115 °C temp. range (50-239 °F)
500 psig pressure
I.D. × O.D.
2.30 mm × 7.75 mm
용적
250 μL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The needles on Thin Layer Chromatography (TLC) syringes are coated from the point for 3/4” with a PTFE-coating. The coating reduces the surface tension between the needle and the liquid making it ideal for reproducible sample spotting. The TLC syringe is available as a cemented needle (SNTLC) or as a replacement needle to be installed into a standard removable needle (RN) syringe. To find the appropriate no needle syringe select general syringes and filter by termination and gauge. Choose the “RN” and “No Needle Included” options to see all syringes that can receive TLC needles. To find the appropriate TLC needle select the TLC option under “Needles” in the main product menu.
법적 정보
Hamilton is a registered trademark of Hamilton Co.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)