주요 문서
NIST1905
Nickel microhardness test block (Knoop)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41116107
NACRES:
NA.24
Grade
certified reference material
Quality Level
포장
pkg of each
제조업체/상표
NIST®
유사한 제품을 찾으십니까? 방문 제품 비교 안내
기타 정보
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Nickel (Ni)
See certificate for values and more details at nist.gov/SRM.
법적 정보
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class Code
11 - Combustible Solids
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)