주요 문서
22470-U
Supelco
Supelco Ferrule Nut Adapter (fits Agilent injectors and non-MS detectors), hexagonal wrenchtight version
pkg of 2 ea
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121815
NACRES:
SB.54
일반 설명
These stainless steel adapters enable you to use general purpose and other 1/16 in. compression ferrules in Agilent GCs. Hexagonal shape allows tightening with a wrench to keep fingers from getting burned.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)