Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

W282155

Sigma-Aldrich

Sweet blood orange oil

greener alternative

natural, product of Simone Gatto

Synonym(s):

Orange oil, Citrus aurantium, Citrus sinensis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2821
MDL number:
UNSPSC Code:
12164502
organoleptic:
fresh; orange; citrus; sweet
grade:
Kosher
natural
biological source:
Citrus sinensis (L.) Osbeck
food allergen:
no known allergens

biological source

Citrus sinensis (L.) Osbeck

Quality Level

grade

Kosher
natural

reg. compliance

FDA 21 CFR 117

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

organoleptic

fresh; orange; citrus; sweet

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

129.2 °F - closed cup

flash_point_c

54 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service