Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01638

Millipore

BROLACIN agar (C.L.E.D. agar)

for the enumeration, isolation and preliminary identification of microorganisms

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41106213

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.2-7.4 (37 °C, 33 g/L in H2O, after autoclaving)

solubility

33.2 g/L

bulk density

570 kg/m3

application(s)

clinical
microbiology

storage temp.

15-25°C

suitability

Escherichia coli
Proteus spp.
Pseudomonas spp.
Salmonella spp.
Shigella spp.
Staphylococcus spp.

General description

for the enumeration, isolation and preliminary identification of microorganisms

Analysis Note

Appearance (clearness)clearAppearance (colour)bluish green to dark greenpH-value (25 °C)7.1 - 7.5Solidification behaviour (2 hrs., 45 °C)liquidRecovery on test medium (Escherichia coli ATCC 11775 (WDCM 00090))≥ 70 %Recovery on test medium (Salmonella typhimurium ATCC 13311 (WDCM 00121))≥ 70 %Recovery on test medium (Shigella flexneri ATCC 29903 (WDCM 00125))≥ 70 %Recovery on test medium (Proteus mirabilis ATCC 29906 (WDCM 00023))≥ 70 %Recovery on test medium (Proteus vulgaris ATCC 8427)≥ 70 %Recovery on test medium (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025))≥ 70 %Recovery on test medium (Staphylococcus aureus ATCC 6538-P (WDCM 00033))≥ 70 %Colony colour (Escherichia coli ATCC 11775 (WDCM 00090))yellowColony colour (Staphylococcus aureus ATCC 6538-P (WDCM 00033))yellowMedium colour (Salmonella typhimurium ATCC 13311 (WDCM 00121))blueMedium colour (Shigella flexneri ATCC 29903 (WDCM 00125))blueMedium colour (Proteus mirabilis ATCC 29906 (WDCM 00023))blueMedium colour (Proteus vulgaris ATCC 8427)blueMedium colour (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025))blueIncubation: 24 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service