Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

394675

Millipore

TAPS

ULTROL® Grade, >98% (titration), zwitterionic buffer useful in pH range 7.7-9.1

Synonym(s):

TAPS, ULTROL® Grade, 3-{[ tris(Hydroxymethyl)methyl]amino}propanesulfonic Acid, 3-{[tris(Hydroxymethyl)methyl]amino}propanesulfonic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO6S
CAS Number:
Molecular Weight:
243.28
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

Quality Level

assay

>98% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (20 °C)

8.4

solubility

water: 200 mg/mL

cation traces

heavy metals: ≤2 ppm

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C7H17NO6S/c9-4-7(5-10,6-11)8-2-1-3-15(12,13)14/h8-11H,1-6H2,(H,12,13,14)

InChI key

YNLCVAQJIKOXER-UHFFFAOYSA-N

General description

A zwitterionic buffer useful in the pH of 7.7-9.1 range. Has a pKa of 8.3-8.5 at 25°C.
A zwitterionic buffer useful in the pH range of 7.7-9.1. Has a pKa of 8.3-8.5 at 25°C.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature. Stock solutions are stable for up to several months at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service