Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX0155

Supelco

Acetonitrile

≥99.9% (GC), suitable for gas chromatography (GC), OmniSolv®, Non UV

Synonym(s):

Acetonitrile, ACN, Cyanomethane, Ethyl nitrile, Methyl Cyanide, Ethanenitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein/REAXYS Number:
741857
MDL number:
UNSPSC Code:
12352117
EC Index Number:
200-835-2
assay:
≥99.9% (GC)
technique(s):
gas chromatography (GC): suitable
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

Product Name

Acetonitrile Non UV, for GC OmniSolv®

vapor density

1.41 (vs air)

Quality Level

vapor pressure

72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

product line

OmniSolv®

assay

≥99.9% (GC)

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

gas chromatography (GC): suitable

impurities

≤0.10% Water (H2O)
≤0.6 μeq/g Titrable base
≤2 ng/L Capillary ECD responsive substances (as PCNB)
≤8.3 μeq/g Titrable acid

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 2 °C

density

0.786 g/mL at 25 °C (lit.)

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

for GC OmniSolv®

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.9% minCapillary ECD responsive substances (as PCNB)2ng/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardTitrable acid8.3µeq/g maxTitrable base0.6µeq/g maxWater (H2O)0.10% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service