Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

936979

Sigma-Aldrich

rac-BINAP Pd G3 ChemBeads

Synonym(s):

[2′-(amino-κN)[1,1′-biphenyl]-2-yl-κC][[2′-(diphenylphosphino)[1,1′-binaphthalen]-2-yl]diphenylphosphine-κP](methanesulfonato-κO)- palladium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H45NO3P2PdS
CAS Number:
Molecular Weight:
992.40
MDL number:
UNSPSC Code:
12352100

form

solid

Quality Level

composition

loading of catalyst, 4-6 wt. %

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

NC1=C(C2=CC=CC=C2[Pd]OS(C)(=O)=O)C=CC=C1.C34=CC=CC=C3C=CC(P(C5=CC=CC=C5)C6=CC=CC=C6)=C4C7=C(P(C8=CC=CC=C8)C9=CC=CC=C9)C=CC%10=C7C=CC=C%10>>

InChI

InChI=1S/C44H32P2.C12H10N.CH4O3S.Pd/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h1-32H;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

BVYJEOTVUJMCBM-UHFFFAOYSA-M

General description

rac-BINAP-Pd-G3 ChemBeads are a third generation (G3) Buchwald precatalyst on glass beads. It is an air, moisture and thermally-stable precatalyst and is highly soluble in a wide range of common organic solvents. It has long life in solutions. rac-BINAP-Pd-G3 ChemBeads are an excellent reagent for palladium catalyzed cross-coupling reactions. Some of thier unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio, relative to earlier generations of Buchwald precatalysts.

Application

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.For general uses, product is also available in powdered form (804967)

Features and Benefits

For feature and benefits, please visit the technical article on Buchwald G3 Precatalysts. https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/g3-and-g4-buchwald-precatalysts

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service