Skip to Content
Merck
All Photos(1)

Key Documents

P216

Sigma-Aldrich

(±)-PD 128,907 hydrochloride

solid, ≥98% (HPLC)

Synonym(s):

trans-(±)-3,4,4a,10b-Tetrahydro-4-propyl-2H,5H-[1]benzopyrano[4,3-b]-1,4-oxazin-9-ol hydrochloride, PD 125,530

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO3 · HCl
CAS Number:
Molecular Weight:
285.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

white

solubility

H2O: >5 mg/mL

SMILES string

Cl.CCCN1CCO[C@H]2[C@H]1COc3ccc(O)cc23

InChI

1S/C14H19NO3.ClH/c1-2-5-15-6-7-17-14-11-8-10(16)3-4-13(11)18-9-12(14)15;/h3-4,8,12,14,16H,2,5-7,9H2,1H3;1H/t12-,14-;/m1./s1

InChI key

DCFXOTRONMKUJB-QMDUSEKHSA-N

Gene Information

human ... DRD3(1814)

Biochem/physiol Actions

Selective D3 dopamine receptor agonist.

Caution

Light sensitive.

Legal Information

Manufactured and sold with the permission of Warner-Lambert Company.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Gobert et al.
Journal of neurochemistry, 66(5), 2209-2212 (1996-05-01)
In freely moving rats, the novel, selective dopamine (DA) D3 receptor agonist PD 128,907 dose-dependently [effective dose (ED25) = 0.07 mg/kg, s.c.] reduced dialysate levels of DA in the frontal cortex, a structure innervated by the ventral tegmental area (VTA).
T A Pugsley et al.
The Journal of pharmacology and experimental therapeutics, 275(3), 1355-1366 (1995-12-01)
The present study determined the biochemical and pharmacological effects of PD 128907 [R-(+)-trans-3,4,4a,10b-tetrahydro-4-propyl-2H,5H- [1]benzopyrano[4,3-b]-1,4-oxazin-9-ol], a dopamine (DA) receptor agonist that shows a preference for the human D3 receptor. In transfected Chinese hamster ovary cells (CHO K1), PD 128907 displaced [3H]spiperone
H A DeWald et al.
Journal of medicinal chemistry, 33(1), 445-450 (1990-01-01)
The dopamine agonist profile of (+-)-trans-3,4,4a,10b-tetrahydro-4-propyl-2H,5H-[1]benzopyrano [4,3-b]-1,4-oxazin-9-ol (16a) and its enantiomers (16b-c) was examined. Racemic 16a exhibited moderate affinity for the dopamine (DA) D2 receptor labeled with the DA antagonist ligand [3H]haloperidol and moderate in vivo activity; it attenuated gamma-butyrolactone-stimulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service