Skip to Content
Merck
All Photos(1)

Key Documents

I0638

Sigma-Aldrich

IPL-41 Insect Medium

suitable for insect cell culture, powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

Pricing and availability is not currently available.

form

powder

Quality Level

technique(s)

cell culture | insect: suitable

storage temp.

2-8°C

Application

IPL-41 is part of a series of media originally developed at the USDA Insect Pathology Laboratory by Weiss et al. for the large scale propagation of cells from the fall armyworm, Spodoptera frugiperda. The medium is primarily used for the growth and maintenance of cell lines derived from lepidopterans and the propagation of viruses in these cell lines. One of the most significant uses of the medium is the large scale culture of baculovirus infected Spodoptera cells. The expression of recombinant proteins is undertaken from Spodoptera cells utilizing the baculovirus expression system (BEVS). Attempts have also been made to obtain the expression of foreign genes in cells cultured in IPL-41 without serum supplementation.

Formula variant

With L-glutamine. Without calcium chloride and sodium bicarbonate.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved method for the production of insect cell cultures in large volumes.
Weiss, S.A., et al.
In Vitro, 17, 495-502 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service