Skip to Content
Merck
All Photos(1)

Key Documents

P0618

Sigma-Aldrich

cis-8-Hydroxy-3-(n-propyl)-1,2,3a,4,5,9b-hexahydro-1H-benz[e]indole hydrobromide

≥98% (HPLC), solid

Synonym(s):

cis(±)-8-OH-PBZI hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO · HBr
Molecular Weight:
312.25
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

assay

≥98% (HPLC)

form

solid

color

light brown

solubility

H2O: soluble ~10 mg/mL at 60 °C (Sonicate)
DMSO: soluble 12 mg/mL

storage temp.

2-8°C

SMILES string

Br.CCCN1CC[C@@H]2[C@H]1CCc3ccc(O)cc23

InChI

1S/C15H21NO.BrH/c1-2-8-16-9-7-13-14-10-12(17)5-3-11(14)4-6-15(13)16;/h3,5,10,13,15,17H,2,4,6-9H2,1H3;1H/t13-,15+;/m0./s1

InChI key

UMGQLEXNQCHIFN-NQQJLSKUSA-N

Gene Information

human ... DRD3(1814)

Biochem/physiol Actions

D3 dopamine receptor agonist.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E V Kuzhikandathil et al.
Pharmaceutical research, 29(8), 2264-2275 (2012-05-02)
The D3 dopamine receptor exhibits tolerance and slow response termination (SRT) properties that are not exhibited by the closely-related D2 dopamine receptor. We previously demonstrated that the induction of tolerance elicits a unique conformational change in the D3 receptor. Here
Ligia Westrich et al.
Biochemical pharmacology, 79(6), 897-907 (2009-11-03)
The D3 but not D2 dopamine receptors exhibit a tolerance property in which agonist-induced D3 receptor response progressively decreases upon repeated agonist stimulation. We have previously shown that the D3 receptor tolerance property is not mediated by receptor internalization, persistent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service