Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0625

Sigma-Aldrich

AC253 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

Amylin Antagonist AC253

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C122H196N40O39 · xC2HF3O2
Molecular Weight:
2847.11 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

assay

≥95% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

Amino Acid Sequence

Ac-Leu-Gly-Arg-Leu-Ser-Gln-Glu-Leu-His-Arg-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Asn-Thr-Tyr-NH2

Biochem/physiol Actions

AC253 is an antagonist of Amylin, one of the peptides in the Calcitonin-gene related peptide (CGRP) family. In recent studies. AC253 restored memories in brain cells taken from animals used as models of Alzheimer′s disease. Application of AC253 significantly enhanced long-term potentiation (LTP) when given to 6- to 12-month-old TgCRND8 mice that normally show blunted LTP. In other studies, AC25 neutralized the depressant effects of Aβ(1-42) on hippocampal LTP without affecting baseline transmission or LTP on its own.

Features and Benefits

This compound is featured on the Calcitonin Gene-Related Peptides page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service