Skip to Content
Merck
All Photos(1)

Key Documents

SRP6320

Sigma-Aldrich

FGF-12 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

FGF12B, FHF1, Fibroblast growth factor 12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

liquid

mol wt

22.6 kDa (201 aa, 1-181 aa + His Tag)

packaging

pkg of 50 μg

concentration

1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FGF12(2257)

General description

FGF12 (Fibroblast growth factor 12) is a member of the FGF superfamily of molecules which currently stands at 22 members. The FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. FGF12 binds to IB2 (islet brain-2), a cellular kinase scaffold, and voltage gated sodium channels and also plays an important role in intracellular signaling and ion exchange. Recombinant human FGF12 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.

Physical form

1 mg/mL in 20 mM Tris pH 7.5, 10% glycerol, 1 mM DTT, 2 mM EDTA.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fibroblast growth factors in cell culture and various growth factors for your research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service