MilliporeSigma
  • Rab6-dependent retrograde traffic of LAT controls immune synapse formation and T cell activation.
Rab6-dependent retrograde traffic of LAT controls immune synapse formation and T cell activation.
The Journal of experimental medicine (2018-02-15)
Jean-Marie Carpier, Andres E Zucchetti, Laurence Bataille, Stéphanie Dogniaux, Massiullah Shafaq-Zadah, Sabine Bardin, Marco Lucchino, Mathieu Maurin, Leonel D Joannas, Joao Gamelas Magalhaes, Ludger Johannes, Thierry Galli, Bruno Goud, Claire Hivroz
ABSTRACT
The adapter molecule linker for activation of T cells (LAT) orchestrates the formation of signalosomes upon T cell receptor (TCR) stimulation. LAT is present in different intracellular pools and is dynamically recruited to the immune synapse upon stimulation. However, the intracellular traffic of LAT and its function in T lymphocyte activation are ill defined. We show herein that LAT, once internalized, transits through the Golgi-trans-Golgi network (TGN), where it is repolarized to the immune synapse. This retrograde transport of LAT depends on the small GTPase Rab6 and the target soluble N-ethylmaleimide-sensitive factor attachment protein receptor (t-SNARE) Syntaxin-16, two regulators of the endosome-to-Golgi/TGN retrograde transport. We also show in vitro in Syntaxin-16- or Rab6-silenced human cells and in vivo in CD4+ T lymphocytes of the Rab6 knockout mouse that this retrograde traffic controls TCR stimulation. These results establish that the retrograde traffic of LAT from the plasma membrane to the Golgi-TGN controls the polarized delivery of LAT at the immune synapse and T lymphocyte activation.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Sigma-Aldrich
Saponin from Quillaja sp., Sapogenin content 20-35 %
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Anti-phospho-LAT (Tyr191) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-LAT Antibody, Upstate®, from rabbit
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
N-Acetyl-D-lactosamine, ≥98% (TLC)
Sigma-Aldrich
Red Blood Cell Lysing Buffer Hybri-Max, liquid, sterile-filtered, suitable for hybridoma