MilliporeSigma
  • Peroral amphotericin B polymer nanoparticles lead to comparable or superior in vivo antifungal activity to that of intravenous Ambisome® or Fungizone™.
Peroral amphotericin B polymer nanoparticles lead to comparable or superior in vivo antifungal activity to that of intravenous Ambisome® or Fungizone™.
PloS one (2011-10-15)
Jagdishbhai L Italia, Andrew Sharp, Katharine C Carter, Peter Warn, M N V Ravi Kumar
ABSTRACT
Despite advances in the treatment, the morbidity and mortality rate associated with invasive aspergillosis remains unacceptably high (70-90%) in immunocompromised patients. Amphotericin B (AMB), a polyene antibiotic with broad spectrum antifungal activity appears to be a choice of treatment but is available only as an intravenous formulation; development of an oral formulation would be beneficial as well as economical. Poly(lactide-co-glycolode) (PLGA) nanoparticles encapsulating AMB (AMB-NPs) were developed for oral administration. The AMB-NPs were 113 ± 20 nm in size with ~70% entrapment efficiency at 30% AMB w/w of polymer. The in vivo therapeutic efficacy of oral AMB-NPs was evaluated in neutropenic murine models of disseminated and invasive pulmonary aspergillosis. AMB-NPs exhibited comparable or superior efficacy to that of Ambisome® or Fungizone™ administered parenterally indicating potential of NPs as carrier for oral delivery. The present investigation describes an efficient way of producing AMB-NPs with higher AMB pay-load and entrapment efficiency employing DMSO as solvent and ethanol as non-solvent. The developed oral formulation was highly efficacious in murine models of disseminated aspergillosis as well as an invasive pulmonary aspergillosis, which is refractory to treatment with IP Fungizone™ and responds only modestly to AmBisome®.
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Reagent Alcohol, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
ECO TWEEN® 80, viscous liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, BioXtra, viscous liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, suitable for cell culture, suitable for insect cell culture, viscous liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, viscous liquid, Low-peroxide
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, for molecular biology, syrup
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, viscous liquid, Preservative Free, Low-peroxide; Low-carbonyls
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, average Mw ~1,380
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, average Mw ~1,460
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, average Mw ~1,540
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, from non-animal source
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethyl alcohol, denatured, reagent grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethanol, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00