MilliporeSigma
  • A versatile microfluidic device for multiple ex vivo/in vitro tissue assays unrestrained from tissue topography.
A versatile microfluidic device for multiple ex vivo/in vitro tissue assays unrestrained from tissue topography.
Microsystems & nanoengineering (2020-06-29)
Jose M de Hoyos-Vega, Alan M Gonzalez-Suarez, Jose L Garcia-Cordero
ABSTRACT
Precision-cut tissue slices are an important in vitro system to study organ function because they preserve most of the native cellular microenvironments of organs, including complex intercellular connections. However, during sample manipulation or slicing, some of the natural surface topology and structure of these tissues is lost or damaged. Here, we introduce a microfluidic platform to perform multiple assays on the surface of a tissue section, unhindered by surface topography. The device consists of a valve on one side and eight open microchannels located on the opposite side, with the tissue section sandwiched between these two structures. When the valve is actuated, eight independent microfluidic channels are formed over a tissue section. This strategy prevents cross-contamination when performing assays and enables parallelization. Using irregular tissues such as an aorta, we conducted multiple in vitro and ex vivo assays on tissue sections, including short-term culturing, a drug toxicity assay, a fluorescence immunohistochemistry staining assay, and an immune cell assay, in which we observed the interaction of neutrophils with lipopolysaccharide (LPS)-stimulated endothelium. Our microfluidic platform can be employed in other disciplines, such as tissue physiology and pathophysiology, morphogenesis, drug toxicity and efficiency, metabolism studies, and diagnostics, enabling the conduction of several assays with a single biopsy sample.
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O128:B12, purified by phenol extraction
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Potassium cyanide, BioUltra, ≥98.0% (AT)