MilliporeSigma
A multi-organ chip with matured tissue niches linked by vascular flow.
Nature biomedical engineering (2022-04-29)
Kacey Ronaldson-Bouchard, Diogo Teles, Keith Yeager, Daniel Naveed Tavakol, Yimu Zhao, Alan Chramiec, Somnath Tagore, Max Summers, Sophia Stylianos, Manuel Tamargo, Busub Marcus Lee, Susan P Halligan, Erbil Hasan Abaci, Zongyou Guo, Joanna Jacków, Alberto Pappalardo, Jerry Shih, Rajesh K Soni, Shivam Sonar, Carrie German, Angela M Christiano, Andrea Califano, Karen K Hirschi, Christopher S Chen, Andrzej Przekwas, Gordana Vunjak-Novakovic
ABSTRACT
Engineered tissues can be used to model human pathophysiology and test the efficacy and safety of drugs. Yet, to model whole-body physiology and systemic diseases, engineered tissues with preserved phenotypes need to physiologically communicate. Here we report the development and applicability of a tissue-chip system in which matured human heart, liver, bone and skin tissue niches are linked by recirculating vascular flow to allow for the recapitulation of interdependent organ functions. Each tissue is cultured in its own optimized environment and is separated from the common vascular flow by a selectively permeable endothelial barrier. The interlinked tissues maintained their molecular, structural and functional phenotypes over 4 weeks of culture, recapitulated the pharmacokinetic and pharmacodynamic profiles of doxorubicin in humans, allowed for the identification of early miRNA biomarkers of cardiotoxicity, and increased the predictive values of clinically observed miRNA responses relative to tissues cultured in isolation and to fluidically interlinked tissues in the absence of endothelial barriers. Vascularly linked and phenotypically stable matured human tissues may facilitate the clinical applicability of tissue chips.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Osteocalcin Antibody, serum, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Collagen Type I, rat tail
Pricing and availability is not currently available.
Sigma-Aldrich
Fibronectin bovine plasma, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Pricing and availability is not currently available.
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280, 18.3)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Cytochrome P450 Enzyme CYP3A4 Antibody, serum, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Bone Sialoprotein II Antibody, serum, Chemicon®
Pricing and availability is not currently available.