MilliporeSigma
  • Improved survival, vascular differentiation and wound healing potential of stem cells co-cultured with endothelial cells.
Improved survival, vascular differentiation and wound healing potential of stem cells co-cultured with endothelial cells.
PloS one (2011-02-02)
Dora C S Pedroso, Ana Tellechea, Liane Moura, Isabel Fidalgo-Carvalho, João Duarte, Eugénia Carvalho, Lino Ferreira
ABSTRACT
In this study, we developed a methodology to improve the survival, vascular differentiation and regenerative potential of umbilical cord blood (UCB)-derived hematopoietic stem cells (CD34(+) cells), by co-culturing the stem cells in a 3D fibrin gel with CD34(+)-derived endothelial cells (ECs). ECs differentiated from CD34(+) cells appear to have superior angiogenic properties to fully differentiated ECs, such as human umbilical vein endothelial cells (HUVECs). Our results indicate that the pro-survival effect of CD34(+)-derived ECs on CD34(+) cells is mediated, at least in part, by bioactive factors released from ECs. This effect likely involves the secretion of novel cytokines, including interleukin-17 (IL-17) and interleukin-10 (IL-10), and the activation of the ERK 1/2 pathway in CD34(+) cells. We also show that the endothelial differentiation of CD34(+) cells in co-culture with CD34(+)-derived ECs is mediated by a combination of soluble and insoluble factors. The regenerative potential of this co-culture system was demonstrated in a chronic wound diabetic animal model. The co-transplantation of CD34(+) cells with CD34(+)-derived ECs improved the wound healing relatively to controls, by decreasing the inflammatory reaction and increasing the neovascularization of the wound.
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Histopaque®-1077 Hybri-Max, liquid, sterile-filtered, BioReagent, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SKU
Pack Size
Availability
Price
Quantity
SAFC
L-Glutamine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)