MilliporeSigma
  • Hypersensitive site 6 of the Th2 locus control region is essential for Th2 cytokine expression.
Hypersensitive site 6 of the Th2 locus control region is essential for Th2 cytokine expression.
Proceedings of the National Academy of Sciences of the United States of America (2013-04-10)
Adam Williams, Gap Ryol Lee, Charalampos G Spilianakis, Soo Seok Hwang, Stephanie C Eisenbarth, Richard A Flavell
ABSTRACT
The T helper type 2 (Th2) cytokine genes Il4, Il5, and Il13 are contained within a 140-kb region of mouse chromosome 11 and their expression is controlled by a locus control region (LCR) embedded within this locus. The LCR is composed of a number of DNase I-hypersensitive sites (HSs), which are believed to encompass the regulatory core of the LCR. To determine the function of these sites, mutant mice were generated in which combinations of these HSs had been deleted from the endogenous LCR, and the effect on Th2 cytokine expression was assessed through the use of in vivo and in vitro models. These experiments revealed that, although all of the hypersensitive sites analyzed are important for appropriate LCR function, some sites are more important than others in regulating cytokine expression. Interestingly, each LCR mutation showed contrasting effects on cytokine expression, in some cases with mutants displaying opposing phenotypes between in vitro cultures and in vivo immunizations. These studies indicated that Rad50 hypersensitive site 6 was the singularly most important HS for Th2 cytokine expression, displaying consistent reductions in cytokine levels in all models tested. Furthermore analysis of chromatin modifications revealed that deletion of Rad50 hypersensitive site 6 impacted epigenetic modifications at the promoters of the Il4, Il5, and Il13 genes as well as other regulatory sites within the Th2 locus.
Sigma-Aldrich
Deoxyribonuclease I RNase-free solution from bovine pancreas
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, powder, 62-88% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, lyophilized powder, RNAse and protease, free
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.