MilliporeSigma
  • Synthesis of N(4)-(substituted phenyl)-N(4)-alkyl/desalkyl-9H-pyrimido[4,5-b]indole-2,4-diamines and identification of new microtubule disrupting compounds that are effective against multidrug resistant cells.
Synthesis of N(4)-(substituted phenyl)-N(4)-alkyl/desalkyl-9H-pyrimido[4,5-b]indole-2,4-diamines and identification of new microtubule disrupting compounds that are effective against multidrug resistant cells.
Bioorganic & medicinal chemistry (2013-01-22)
Aleem Gangjee, Nilesh Zaware, Ravi Kumar Vyas Devambatla, Sudhir Raghavan, Cara D Westbrook, Nicholas F Dybdal-Hargreaves, Ernest Hamel, Susan L Mooberry
ABSTRACT
A series of fourteen N(4)-(substituted phenyl)-N(4)-alkyl/desalkyl-9H-pyrimido[4,5-b]indole-2,4-diamines was synthesized as potential microtubule targeting agents. The synthesis involved a Fisher indole cyclization of 2-amino-6-hydrazinylpyrimidin-4(3H)-one with cyclohexanone, followed by oxidation, chlorination and displacement with appropriate anilines. Compounds 6, 14 and 15 had low nanomolar potency against MDA-MB-435 tumor cells and depolymerized microtubules. Compound 6 additionally had nanomolar GI(50) values against 57 of the NCI 60-tumor panel cell lines. Mechanistic studies showed that 6 inhibited tubulin polymerization and [(3)H]colchicine binding to tubulin. The most potent compounds were all effective in cells expressing P-glycoprotein or the βIII isotype of tubulin, which have been associated with clinical drug resistance. Modeling studies provided the potential interactions of 6, 14 and 15 within the colchicine site.
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Indole, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Supelco
Aniline, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Indole, ≥99%, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Aniline hydrochloride, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Supelco
Cyclohexanone, Selectophore, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Cyclohexanone, 99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Supelco
Cyclohexanone, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Cyclohexanone, SAJ first grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Cyclohexanone, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Cyclohexanone, ReagentPlus®, 99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00