MilliporeSigma
  • Solution growth of spherulitic rod and platelet calcium phosphate assemblies through polymer-assisted mesoscopic transformations.
Solution growth of spherulitic rod and platelet calcium phosphate assemblies through polymer-assisted mesoscopic transformations.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Vassiliki A Kosma, Konstantinos G Beltsios
ABSTRACT
Solution growth of apatite its precursors in the presence of urea commercial gelatin is found to lead, under appropriate conditions, to a rich spectrum of morphologies, among them high aspect ratio needles in uniform sturdy spherulitic assemblies resulting from a herein documented morphological 'Chrysalis Transformation'; the latter transformation involves the growth of parallel arrays of high aspect ratio needles within micron-scale tablets the formation of a radial needle arrangement upon disruption of tablet wrapping. A different level of gelatin leads to the formation of sturdy platelet-based spherulites through another morphological transformation. We also probe the role of four simple synthetic water-soluble polymers; we find that three of them (poly(vinyl alcohol), polyvinylpyrrolidone and polyacrylamide)) also affect substantially the assembly habits of apatite; the effect is similar to that of gelatin but the attained control is less perfect/complete. The case of poly(vinyl alcohol) provides, through variation of the degree of hydrolysis, insights as regards the chain architecture features that might favor morphological transformations. Morphological transformations of particle assemblies documented herein constitute novel ways of generating dense quasi-isotropic reinforcements with high aspect ratio ceramic particles; it becomes possible to tailor calcium phosphate phases at the structural level of crystal assembly.
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, Molecular Biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Sigma-Aldrich
Polyacrylamide, average Mn 150,000