MilliporeSigma
  • First evidence of sucrose biosynthesis by single cyanobacterial bimodular proteins.
First evidence of sucrose biosynthesis by single cyanobacterial bimodular proteins.
FEBS letters (2013-04-27)
Giselle M A Martínez-Noël, Andrea C Cumino, María de los Angeles Kolman, Graciela L Salerno
ABSTRACT
The net synthesis of sucrose (Suc) is catalysed by the sequential action of Suc-phosphate synthase (SPS) and Suc-phosphate phosphatase (SPP). SPS and SPP from Anabaena sp. PCC 7120 (7120-SPS and 7120-SPP) define minimal catalytic units. Bidomainal SPSs, where both units are fused, occur in plants and cyanobacteria, but they display only SPS activity. Using recombinant proteins that have fused 7120-SPS and 7120-SPP, we demonstrated that they are bifunctional chimeras and that the arrangement 7120-SPS/SPP is the most efficient to catalyse the sequential reactions to yield Suc. Moreover, we present the first evidence of a bidomainal SPS present in the cyanobacterium Synechococcus elongatus PCC 7942 with both, SPS and SPP activity.
Product Description
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, SAJ first grade
Sigma-Aldrich
Sucrose, JIS special grade
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sucrose, European Pharmacopoeia (EP) Reference Standard