MilliporeSigma
Bacteria detected on surfaces of formalin fixed anatomy cadavers.
Italian journal of anatomy and embryology = Archivio italiano di anatomia ed embriologia (2013-08-01)
Burton Tabaac, Geoffrey Goldberg, Lia Alvarez, Molly Amin, Kathleen Shupe-Ricksecker, Fernando Gomez
ABSTRACT
The purpose of this study is to determine if anatomy cadavers fixed in a formalin solution are a possible source of introduction of microorganisms into the anatomy laboratory. Routinely preserved cadavers were sampled for microbiological contaminates prior to examination and dissection by anatomy students. Regions sampled include the axilla, oral/nasal cavity, and inguinal/perineal region. Using conventional bacteriologic culture and identification methods our research group was able to successfully recover and identify a variety of organisms from all cadavers and in all regions tested. The results indicate that cadavers processed with 10% buffered formalin have viable organisms on their surfaces that can be a source of contamination of laboratory equipment and clothing. Given the diversity of bacterial species cultured, preserved cadavers used for anatomy education as well as research must be considered a possible source for dissemination of bacterial organisms. This study underscores the importance of standard infection control protocols.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O